Product Image
Thermo Scientific™

Triglycerides Reagent

Determine quantity of triglycerides in human serum or plasma in vitro with Thermo Scientific™ Triglycerides Reagent.
Have Questions?
Número do catálogoQuantity
TR224212 x 125 mL
Número do catálogo TR22421
Preço (BRL)
-
Quantity:
2 x 125 mL
Determine quantity of triglycerides in human serum or plasma in vitro with Thermo Scientific™ Triglycerides Reagent.
Measurement of triglycerides is important in the diagnosis and management of hyperlipidaemias. Levels may be increased in nephrotic syndrome, coronary artery disease (CAD), hypothyroidism, diabetes mellitus and liver disease. Decreased values are sometimes observed in cases of protein malnutrition, hyperthyroidism and cachectic states.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Especificações
DescriptionInfinity Triglyceride 2 x 125mL
LinearityUp to 10mmol/L (885mg/dL)
Methods CompatibilityEndpoint
Quantity2 x 125 mL
Sample TypePlasma, Serum
Stability18 months at 2-8°C
TypeReagent
Detectable AnalytesTriglycerides
Unit SizeEach