Direct Bilirubin Reagent
Thermo Scientific™

Direct Bilirubin Reagent

Determine the quantity of direct Bilirubin in serum with this reagent.
Have Questions?
Catalog NumberQuantity
TR333212 x 125 mL
Catalog number TR33321
Price (MXN)
-
Quantity:
2 x 125 mL
Determine the quantity of direct bilirubin in serum in vitro with Thermo Scientific™ Direct Bilirubin Reagent.
Features:
  • Utilizes the acid diazo method
  • Offers at least 21 days working reagent stability
  • Suitable for manual use or on automated clinical chemistry analyzers
  • Applications for a wide range of automated analyzers available on request
Notes:
In the liver, bilirubin is conjugated with glucuronic acid to form conjugated bilirubin. Increased conjugated or direct bilirubin generally results from acute hepatitis or when any portion of the biliary tree becomes blocked or abnormally permeable.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionBilirubin
Linearity0 to 20mg/dL
Methods CompatibilityEndpoint
Quantity2 x 125 mL
Sample TypeSerum
Stability21 days (Working)
State at Room TemperatureLiquid
TypeReagent
CE MarkerYes
Detectable AnalytesBilirubin
FormLiquid
Unit SizeEach
Contents & Storage
2°C to 8°C