Product Image
Thermo Scientific™

Albumin Reagent

Determine in vitro quantity of albumin in serum or plasma.
Have Questions?
Catalog NumberQuantity
TR360262 x 250 mL
Catalog number TR36026
Price (MXN)
-
Quantity:
2 x 250 mL
Determine the quantity of albumin in serum or plasma in vitro with Thermo Scientific™ Albumin Reagent.
Features:
  • Available in various configurations and kit sizes to suit small and large laboratory needs
  • Suitable for manual use or on automated clinical chemistry analyzers
  • Applications for a wide range of automated analyzers available on request
  • Utilizes the Bromocresol Green substrate
Notes:
Albumin is the most abundant protein in human plasma, representing 55 to 65% of the total protein. Albumin has a relatively small size which makes it a useful indicator of the integrity of glomerular and other membranes. Hyperalbumin is associated with dehydration while hypoalbuminemia is associated with many illnesses including, but not limited to, kidney disease, liver disease, tissue damage, inflammation and malnutrition.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionAlbumin - Bromocresol Green
Linearity0 to 60g/L
Methods CompatibilityEndpoint
Quantity2 x 250 mL
Sample TypePlasma, Serum
State at Room TemperatureLiquid
TypeReagent
CE MarkerYes
Detectable AnalytesAlbumin, Albumin
FormLiquid
Unit SizeEach
Contents & Storage
2°C to 8°C