Thermo Scientific™
Microprotein Reagent
Catalog number: TR50001
Thermo Scientific™
Microprotein Reagent
Catalog number: TR50001
Test for protein in urine and cerebrospinal fluid with Thermo Scientific™ Microprotein Reagent, a stable liquid that minimizes laboratory reagent waste.
Catalog Number
TR50001
Unit Size
Each
Description
Microprotein - Benzethonium Chloride (2-Part Rgts)
Price (USD)
Full specifications
Certifications/ComplianceCE Marked
Quantity1 x 125 mL (R1), 1 x 25 mL (R2)
CE MarkerYes
DescriptionMicroprotein - Benzethonium Chloride (2-Part Rgts)
Detectable AnalytesMicroprotein
Linearity5 to 200mg/dL
Methods CompatibilityTurbidimetric
StabilityStable Liquid
State at Room TemperatureLiquid
FormatLiquid
Sample TypeCSF, Urine
TypeControl
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
TR50001Full specifications
EachMicroprotein - Benzethonium Chloride (2-Part Rgts)Request A Quote
Certifications/ComplianceCE Marked
Quantity1 x 125 mL (R1), 1 x 25 mL (R2)
CE MarkerYes
DescriptionMicroprotein - Benzethonium Chloride (2-Part Rgts)
Detectable AnalytesMicroprotein
Linearity5 to 200mg/dL
Methods CompatibilityTurbidimetric
StabilityStable Liquid
State at Room TemperatureLiquid
FormatLiquid
Sample TypeCSF, Urine
TypeControl
Unit SizeEach
Showing 1 of 1
Protein may be present in the urine under non-pathological conditions such as pregnancy, dehydration and exercise. Pathologic increases are seen with severe burns, nephrotic syndrome, multiple myeloma, heavy metal poisoning, diabetic nephropathy, pyelonephritis and preeclampsia. Protein will be elevated in cerebrospinal fluid with meningitis, encephalitis, brain tumors, intracranial hemorrhage, traumatic injury and poliomyelitis.
  • Intended for use with both manual and automated systems
For In Vitro Diagnostic Use.  
Contents & Storage
2°C to 25°C
Figures