Product Image
Thermo Scientific™
Clinical Chemistry Standards
Perform in vitro diagnosis with ammonia, HDL cholesterol and lithium clinical chemistry standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TR609016 x 5 mL
TR669011 x 10 mL
Catalog number TR60901
Price (MXN)
-
Quantity:
6 x 5 mL
Perform in vitro diagnosis with Thermo Scientific™ Clinical Chemistry Standards. Product include standards for ammonia, HDL cholesterol and lithium. All standards are manufactured to exacting tolerances and, wherever possible, calibrated against National Institute of Standards and Technology (NIST™) materials.
  • Most standards bottled with tapered dropper tips to guard against contamination and evaporation losses during handling and storage
  • For in vitro diagnostic use only
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration100, 200, 300 μg/dL
DescriptionClinical Chemistry Standard calibrated against NIST(National Institute of Standards and Technology) materials
Quantity6 x 5 mL
TypeControl, Standards
Detectable AnalytesAmmonia, HDL Cholesterol, Lithium
Sample TypePlasma, Serum, Urine
Unit SizeEach
Contents & Storage
10°C to 25°C