Thermo Scientific™

AST/GOT Reagent

Catalog number: TR70121
Thermo Scientific™

AST/GOT Reagent

Catalog number: TR70121
Determine quantity of aspartate aminotransferase in serum or plasma in vitro with Thermo Scientific™ AST/GOT, featuring a 30-day reconstituted stability.
 
Catalog Number
TR70121
Unit Size
Each
Description
Infinity AST/GOT
Quantity
2 x 125 mL
Price (USD)
Full specifications
Certifications/ComplianceCE Marked
DescriptionInfinity AST/GOT
CE MarkerYes
Detectable AnalytesAST/GOT
FormatLiquid
Sample TypePlasma, Serum
TypeReagent
Linearity0 to 450U/L
Methods CompatibilityKinetic UV
Quantity2 x 125 mL
StabilityStable Liquid
State at Room TemperatureLiquid
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionQuantityPrice (USD)
TR70121Full specifications
EachInfinity AST/GOT2 x 125 mLRequest A Quote
Certifications/ComplianceCE Marked
DescriptionInfinity AST/GOT
CE MarkerYes
Detectable AnalytesAST/GOT
FormatLiquid
Sample TypePlasma, Serum
TypeReagent
Linearity0 to 450U/L
Methods CompatibilityKinetic UV
Quantity2 x 125 mL
StabilityStable Liquid
State at Room TemperatureLiquid
Unit SizeEach
Showing 1 of 1

AST is widely distributed with high concentrations in the heart, liver, skeletal muscle, and kidney. Damage or disease to any of these tissues such as myocardial infarction, viral hepatitis, liver necrosis, cirrhosis and muscular dystrophy may result in raised serum levels of AST.

  • Reagent is suitable for manual use or on automated clinical chemistry analyzers
  • Applications for a wide range of automated analyzers are available on request.
For In Vitro Diagnostic Use.  

Contents & Storage

2°C to 8°C

Figures