Certifications/Compliance | CE Marked |
Quantity | 2 x 15 mL |
CE Marker | Yes |
Description | Infinity ALT/GPT, Liquid |
Detectable Analytes | ALT/GPT |
Linearity | 0 to 1500U/L |
Methods Compatibility | Kinetic UV |
Stability | Stable Liquid |
State at Room Temperature | Liquid |
Format | Liquid |
Sample Type | Serum |
Type | Reagent |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TR71121 | Each | Infinity ALT/GPT, Liquid | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|