Thermo Scientific™
Lab Vision™ Streptavidin Alkaline Phosphatase (Ready-To-Use)
 Related applications:
Anatomical Pathology
Incubate slides for 10 minutes with Thermo Scientific™ Lab Vision™ Streptavidin Alkaline Phospatase (Ready-to-Use) reagent for results that will remain stable for 18 months. Reagent is intended for use in a labeled streptavidin-biotin immunoenzymatic antigen detection system.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Storage Conditions: Store at 2°-8°C. Each component is stable for 18 months.
STAINING PROTOCOL (kit components in bold):
1. Deparaffinize and rehydrate tissue section.
2. Wash 2 times in buffer.
3. If required, incubate tissue in digestive enzyme (or appropriate pretreatment).
4. Wash 4 times in buffer.
5. (Optional) Place slide in Ultra V Block and incubate for 5-10 minutes at room temperature to block nonspecific background staining. NOTE: Do not exceed 10 minutes or there may be a reduction in desired stain.
6. Rinse (Optional).
7. Apply primary antibody and incubate according to manufacturer's protocol.
8. Wash 4 times in buffer.
9. Incubate slide with biotinylated secondary solution according to manufacturer's protocol.
10. Wash 4 times in buffer.
11. Incubate slide with Streptavidin Alkaline Phosphatase solution for 10 minutes at room temperature.
12. Rinse 4 times in buffer.
13. Place slide in appropriate chromogenic substrate and incubate until desired reaction is achieved.
14. Counterstain and cover slip.
NOTE: This reagent is provided in bulk form for use with automated systems or in staining jars for repeated use. If the DIP method is employed, staining jar should be sealed and refrigerated between uses. Remove staining jars from the refrigerator 30 minutes prior to staining to allow reagents to come to room temperature. The specificity and sensitivity of antigen detection is dependent on the specific primary antibody used.
Documents
Manuals & protocols