MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent
MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent
Thermo Scientific™

MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStandard Components
TS-48920MTBSTFA
TS-48927MTBSTFA + 1% TBMDS
Catalog number TS-48920
Price (USD)
-
Request A Quote
Standard Components:
MTBSTFA

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent. MTBSTFA derivatizes hydroxyl, carboxyl, thiol and primary and secondary amines with typically >96% yields, and the reaction byproducts are volatile and neutral. TBDMS ethers are 104 times more stable to hydrolysis than the corresponding TMS ethers. Addition of Thermo Scientific™ TBDMCS increases silylation potential.

Additional Product Information:
  •  Derivatives have a high molecular concentration at M-57.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
HazardsIrritant
Storage2°C to 8°C
For Use WithGC
Quantity5 mL ampules
Standard ComponentsMTBSTFA
Standard Concentration98 to 100%
Unit SizeEach