Amies Agar Gel Mini Tip: Firm Applicator Wire
Amies Agar Gel Mini Tip: Firm Applicator Wire
Thermo Scientific™

Amies Agar Gel Mini Tip: Firm Applicator Wire

Simplify the transportation of eye, nasopharyngeal, male urethral swab and pediatric swabs with these Amies Agar Gel Transport Swabs.
Have Questions?
Catalog NumberQuantity
TS0012A50/Pk., 500/Cs.
Catalog number TS0012A
Price (MXN)
-
Quantity:
50/Pk., 500/Cs.

Simplify the transportation of eye, nasopharyngeal, male urethral swab and pediatric sampling with Thermo Scientific™ Oxoid™ Amies Agar Gel Mini Tip: Firm Applicator Wire. The narrow dimension of the swab shaft and tip provides a more practical device for some sampling.

Ensure optimal sample collection and recovery using Amies Agar Gel Mini Tip: Firm Applicator Wire.

  • Contains media for the maintenance of bacterial sample during transport to the laboratory.
  • Each batch of product is tested for performance using a wide range of aerobic and anaerobic pathogens to ensure adequate recoveries.
  • Lot number/expiry date and tamper-evident tube ensure complete traceability and sterility.
  • 'Venturi' Hour-Glass Tube Design prevents disintegration of gel column and eliminates undesirable air pockets, bubbles and breaks in gel which may be harmful to fastidious bacteria

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionAmies Agar Gel Mini Tip: Firm Applicator Wire
Quantity50/Pk., 500/Cs.
Shaft MaterialAluminum Wire
Storage Requirements5°C to 25C°; do not freeze
Tip MaterialRayon
Tip TypeMinitip
Product TypeTransport Swab
Unit SizeEach