Direct TIBC Reagents
Product Image
Thermo Scientific™

Direct TIBC Reagents

Decrease turnaround time with Thermo Scientific™ Direct TIBC Reagents, which deliver a fully automated direct measurement of total iron-binding capacity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
TS3000Direct TIBC Calibrator
TS3500Direct TIBC Reagent
Catalog number TS3000
Price (USD)
-
Description:
Direct TIBC Calibrator
Decrease turnaround time with Thermo Scientific™ Direct TIBC Reagents, which deliver a fully automated direct measurement of total iron-binding capacity.
This assay streamlines laboratory testing by maximizing lab productivity, minimizing operator error, improving turnaround times and eliminating the need for sample splitting. From centrifuge to analyzer, no sample pre-treatment is necessary.
  • Increase accuracy of results by as much as 20% over UIBC+ Iron methodologies1
  • Excellent correlation with Alumina Column, Magnetic and Transferrin TIBC methodologies
  • Unaffected by lipemia, bilirubinemia, hemolysis and other potentially interfering compounds
  • Liquid stable, ready-to-use reagent
  • Applications available for a variety of chemistry analyzers
1Siek G, Lawlor J, Pelczar D, Sane M, Musto J. Direct Serum Total Iron-binding Capacity Assay Suitable for Automated Analyzers. Clinical Chemistry. 2002; 48(1):161-166
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceNot CE Marked
DescriptionDirect TIBC Calibrator
Methods CompatibilityColorimetric/Endpoint
Quantity6 x 1 mL
Sample TypeSerum
Stability18 months
State at Room TemperatureLiquid
TypeCalibrator
CE MarkerNo
Detectable AnalytesIron
FormLiquid
Unit SizeEach
Contents & Storage
2°C to 8°C