TSC Series -40°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.4L) capacity, 115V/60Hz, 6.5A - FAQs
View additional product information for TSC Series -40°C Ultra-Low Temperature Chest Freezers - FAQs (TSC1750A)