Precision™ Circulating Water Baths
Thermo Scientific™

Precision™ Circulating Water Baths

Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on these circulating water baths. Featuring advanced temperature controller.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Model
TSCIR3535 LPrecision CIR 35
TSCIR1919 LPrecision CIR 19
TSCIR8989 LPrecision CIR 89
Catalog number TSCIR35
Price (USD)
4,054.12
Online Exclusive
4,610.00
Save 555.88 (12%)
Each
Capacity (Metric):
35 L
Model:
Precision CIR 35
Price (USD)
4,054.12
Online Exclusive
4,610.00
Save 555.88 (12%)
Each

Ensure temperature uniformity and control with theseThermo Scientific™ Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.

  • Achieve enhanced temperature uniformity with perimeter-directed water flow
  • Easily clean and maintain bath with coil-free internal design
  • Optimize scheduling with auto-on and auto-off timers
  • Accommodate taller labware with new hinged lid and extended height
  • Help prevent bath overheating and damage with low-fluid protection
  • Easily operate and monitor with icon-based graphical display
  • Protect your work with audible alarms
Not for use for IVD/MD in some countries.
Specifications
Amperage11.2-13.1 / 5.4-6.3 (120V/230V)
Capacity (English)9.2 gal.
Certifications/ComplianceUL, CE
CommunicationsDigital
Controller TypeDigital
Dimensions (D x W x H) Interior12 x 27.3 x 7.6 in. (305 x 692 x 192 mm)
Dimensions (L x W x H)15.5 x 36.9 x 9.8 in. (394 x 938 x 249 mm)
DisplayMonochrome LCD
Heating Capacity1500 w
Includes1 Stainless Steel Gable cover and Rubber Duck
Material304 Stainless Steel
ModelPrecision CIR 35
Temperature Presets4
Temperature Stability±0.1°C
Temperature Uniformity±0.05°C
Capacity (Metric)35 L
Electrical RequirementsGlobal Voltage 100/115 V 200/230 V 50/60 Hz
Temperature RangeAmbient +5° to 100°C
Unit SizeEach