CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
Thermo Scientific™

CryoMed™ Controlled-Rate Freezer, General Purpose

Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Electrical RequirementsModel
TSCM17PL17 L100 V, 50/60HzGeneral Purpose with Printer
TSCM17PA17 L120 V, 60HzGeneral Purpose with Printer
TSCM17PV17 L220-230 V, 50/60HzGeneral Purpose with Printer
TSCM34PA34 L120 V, 60HzGeneral Purpose with Printer
TSCM34PL34 L100 V, 50/60HzGeneral Purpose with Printer
TSCM34PV34 L220-230 V, 50/60HzGeneral Purpose with Printer
TSCM48PA48 L120 V, 60HzGeneral Purpose with Printer
TSCM48PL48 L100 V, 50/60HzGeneral Purpose with Printer
TSCM48PV48 L220-230 V, 50/60HzGeneral Purpose with Printer
TSCM17XA17 L120 V, 60HzGeneral Purpose without Printer
TSCM17XL17 L100 V, 50/60HzGeneral Purpose without Printer
TSCM17XV17 L220-230 V, 50/60HzGeneral Purpose without Printer
TSCM34XA34 L120 V, 60HzGeneral Purpose without Printer
TSCM34XL34 L100 V, 50/60HzGeneral Purpose without Printer
TSCM34XV34 L220-230 V, 50/60HzGeneral Purpose without Printer
TSCM48XA48 L120 V, 60HzGeneral Purpose without Printer
TSCM48XL48 L100 V, 50/60HzGeneral Purpose without Printer
TSCM48XV48 L220-230 V, 50/60HzGeneral Purpose without Printer
Catalog number TSCM17PL
Price (MXN)
-
Capacity (Metric):
17 L
Electrical Requirements:
100 V, 50/60Hz
Model:
General Purpose with Printer
For customers in research, cell and gene therapy, vaccine production, and biobanking, the CryoMed Controlled-Rate Freezer (CRF) provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.

Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.

User-friendly operation with enhanced data traceability

  • Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
  • Six pre-set freezing profiles and space for up to 14 user-defined, “custom” freeze profiles
  • Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
  • USB data export of .pdf file run logs support 21 CFR part 11
  • User security: 3 levels of user accessibility
  • “Run last” feature allows the same profile to be run consecutively with the touch of a button
  • PC interface software provides remote operation capabilities, run review, and custom profile creation

Real-time run monitoring for sample protection

  • Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing “real time” responsiveness
  • Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
  • Remote alarm contacts
  • Optional thermal printer

Reliable temperature performance for high throughput usage

  • Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
  • Preventative maintenance indicator for LN2 solenoid replacement limits downtime
  • Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
  • Compatible with VHP cleaning techniques
  • Environmentally friendly insulating foam
  • Type 304 stainless steel with exterior powder-coat finish

Warranty varies by region. Check with your local representative for more information

Specifications
Amperage12 A
ConnectivityThermo Fisher Connect compatible
Data OutputsUSB data export
Dimensions (DxWxH) Exterior24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)
DisplayTouchscreen User Interface
Electrical Requirements100 V, 50/60Hz
Exterior FinishPowder-coated Steel
ModelGeneral Purpose with Printer
On-Board DataloggingYes
PC ProgrammabilityPC Interface Software Supported
Sample LoadingFront-loading
TypeControlled-rate freezer
Vial Capacity<380
WarrantyWarranty varies by region. Contact your local sales representative for more information
Weight (English)154 lb.
Weight (Metric)69.8 kg
Cabinet MaterialStainless Steel
Capacity (English)0.6 cu. ft.
Capacity (Metric)17 L
Certifications/CompliancecULus
Plug TypeNEMA 5-15 P
Temperature Range-180°C to +50°C
Unit SizeEach