TSD Series -40°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9L) capcity, holds 320 boxes, 230V/50Hz, 12A - FAQs
TSD Series -40°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9L) capcity, holds 320 boxes, 230V/50Hz, 12A - FAQs
View additional product information for TSD Series -40°C Upright Ultra-Low Temperature Freezers - FAQs (TSD40320V)