TSHP FMS High-Performance Laboratory Freezer, -20°C; 23 cu. ft., 115V
Thermo Scientific™

TSHP FMS High-Performance Laboratory Freezer, -20°C; 23 cu. ft., 115V

Protect flammable materials, solvents and intermediates using Thermo Scientific™ TSHP FMS High-Performance Laboratory Freezers.
Have Questions?
Número de catálogoCapacidad (imperial)Capacidad (métrico)Tensión
TSFMS2320A23 cu. ft.651 L115 V
Número de catálogo TSFMS2320A
Precio (MXN)
-
Capacidad (imperial):
23 cu. ft.
Capacidad (métrico):
651 L
Tensión:
115 V

Protect flammable materials, solvents and intermediates using Thermo Scientific™ TSHP FMS High-Performance Laboratory Freezers, which meet stringent U.S. Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids.† These freezers are designed to maintain a safe environment while keeping valuable samples within a tight temperature range. Each is listed by UL as a special purpose freezer intended for flammable material storage in accordance with NFPA 45, 70 and 99.† 45, 70 and 99.†

Key Features
  • Easy-to-use, intuitive digital display featuring microprocessor control
  • Lockable doors, audio and visual alarms including door ajar alarms for security and peace of mind
  • Convenient access port and casters
  • Outstanding construction featuring painted, high-quality steel exteriors with durable finish
  • Ignition-free interior cabinet design for the safe storage of flammable materials
  • Meets safety standards in accordance with UL, NFPA† and OSHA† for regulatory compliance
  • Optional, built-in, inkless, seven-day graphic chart recorder
  • Optional, barrier-protected 4-20 milliamp output for external monitoring
  • Manual defrost

Ordering Information

Temperature chart recorders, and 4-20 milliamp are available separately. Not Available Outside North America.

Products complying with the applicable requirements of NFPA 70, "National Electric Code", NPFA 45, "Fire Protection for Laboratories Using Chemicals," and NFPA 99 "Health Care Facilities" are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.
Especificaciones
DescongelaciónManual
Fondo exterior (imperial)37 in.
Fondo interior (imperial)29 in.
Fondo exterior (métrico)94 cm
Fondo interior (métrico)73.7 cm
Dimensiones interiores (F x An x Al)29 x 24 x 58 in. (73.7 x 61 x 147.3 cm)
Requisitos eléctricos115 V 60 Hz
Frecuencia60 Hz
Altura exterior (imperial)78.6 in.
Altura (imperial) interior58 in.
Altura exterior (métrico)200 cm
Altura (métrico) interior147.3 cm
AislamientoCFC-free
RefrigeranteCFC-free
Anchura exterior (imperial)28 in.
Anchura (imperial) interior24 in.
Anchura exterior (métrico)72 cm
Anchura (métrico) interior61 cm
Capacidad (imperial)23 cu. ft.
Capacidad (métrico)651 L
Certificaciones/conformidadUL
Opciones de vigilanciaExternal
No. of Doors1
Estantes4
Intervalo de temperatura-20°C
Tensión115 V
Unit SizeEach