TSG General Purpose Freezers - Boxed
Thermo Scientific™

TSG General Purpose Freezers - Boxed

The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Plug Type
TSG1500FESBV1500 LEU
TSG1500FESBW1500 LUK
TSG400FESBV400 LEU
TSG400FESBW400 LUK
TSG700FESBV700 LEU
TSG700FESBW700 LUK
Catalog number TSG1500FESBV
Price (MXN)
-
Capacity (Metric):
1500 L
Plug Type:
EU
The Thermo Scientific™ TSG Series general purpose laboratory freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series Freezers are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from solid door 400 L to 1500 L double door.
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
Specifications
Cabinet MaterialSteel
Capacity (Metric) Shelf30 kg
CastersYes
Control TypeTouchscreen Control with Password Protection
DefrostAdaptive Defrost
DescriptionTSG Series General Purpose Laboratory Freezer
Dimensions (D x W x H) Interior690 x 1300 x 1510 mm
Dimensions (L x W x H) Exterior837 x 1440 x 2082 mm
Door StyleSolid
Electrical Requirements230V/50Hz
Frequency50 Hz
InteriorPainted Steel
Packaging TypeBoxed
Plug TypeEU
Ports1 Rear Access Port
RefrigerantR290
Shipping Weight (Metric)263 kg
Smart View CompatibilitySmart-Vue Pro Compatible
Temperature (Setpoint)-20°C
Warranty36 Months
Capacity (Metric)1500 L
Certifications/ComplianceCE
Monitoring OptionsAlarm
No. of Doors2
Shelves8 Adjustable
Temperature Range-12°C to -30°C
Voltage230 V
Unit SizeEach