TSG Series Solid Door Laboratory Refrigerators
TSG Series Solid Door Laboratory Refrigerators
TSG Series Solid Door Laboratory Refrigerators
TSG Series Solid Door Laboratory Refrigerators
Thermo Scientific™

TSG Series Solid Door Laboratory Refrigerators

Green features
The TSG Solid Door Refrigerators offer excellent performance for sample storage needing 2-8 ° C, an outstanding 4-year parts and labor warranty, and are available in single and double door configurations. All TSG Series Refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigeration.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)InteriorElectrical Requirements
TSG2305SA23 cu. ft.White (Painted)115V/60Hz
TSG1205SA12 cu. ft.White (Painted)115V/60Hz
TSG1205SD12 cu. ft.White (Painted)208-230V/60Hz
TSG2305SD23 cu. ft.White (Painted)208-230V/60Hz
TSG3005SA29.2 cu. ft.White (Painted)115V/60Hz
TSG3005SD29.2 cu. ft.White (Painted)208-230V/60Hz
TSG5005SA51.1 cu. ft.White (Painted)115V/60Hz
TSG5005SD51.1 cu. ft.White (Painted)208-230V/60Hz
Catalog number TSG2305SA
Price (MXN)
-
Capacity (English):
23 cu. ft.
Interior:
White (Painted)
Electrical Requirements:
115V/60Hz
Thermo Scientific TSG Series Solid Door Laboratory Refrigerators offer quiet, intuitive operation with excellent temperature performance and an outstanding warranty, meeting the needs of a wide variety of professional users who need consistent, reliable performance without sacrificing usability or sustainability. All TSG Series Laboratory Refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigerants and are configured to provide excellent temperature performance while minimizing power consumption, all delivered from our zero waste to landfill facility in Asheville, North Carolina.

The drive for sample integrity and security

  • Designed to safely store media, reagents, and other laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned solid doors with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Extra and Alternate wire baskets, shelves, and drawers
  • Stainless Steel interior finish available
  • E-Lock Adapter kits
Specifications
Access SecurityKey Lock
Cabinet MaterialPainted Steel
Capacity (English) Shelf100 lb.
Capacity (Metric) Shelf45 kg
Control TypeCapacitive Touch
DescriptionTSG Series General Purpose Refrigerator
Dimensions (DxWxH) Exterior37.9 x 28.0 x 78.5 in. (962 x 711 x 1994 mm)
Dimensions (D x W x H) Interior28.5 x 24 x 58 in. (723 x 610 x 1473 mm)
Electrical Requirements115V/60Hz
Exterior FinishPainted
Green FeaturesENERGY STAR
InteriorWhite (Painted)
Monitoring OptionsSmartVue Pro, Remote Alarm Contacts
Ports1 Rear Access Port
RefrigerantR290
Shelves4 Adjustable
Shipping Weight (English)426 lb.
Shipping Weight (Metric)193 kg
Temperature (Setpoint)+5°C
TypeGeneral Purpose Refrigerator
Warranty4 Years Parts and Labor + 1 Year Compressor
Capacity (English)23 cu. ft.
Capacity (Metric)650 L
Certifications/CompliancecULus
CompressorHermetic
Door StyleSolid
No. of Doors1
Plug TypeNEMA 5-15
StyleUpright
Temperature Range3°C to 7°C
Voltage115 V
Unit SizeEach