TSG Series Glass Door Laboratory Refrigerators
TSG Series Glass Door Laboratory Refrigerators
TSG Series Glass Door Laboratory Refrigerators
TSG Series Glass Door Laboratory Refrigerators
Thermo Scientific™

TSG Series Glass Door Laboratory Refrigerators

The TSG Glass Door Refrigerators offer sustainability at 2-8°C, an outstanding 4-year parts and labor warranty, and tight temperature uniformity in single and double door configurations. All TSG Series utilize SNAP and F-Gas compliant refrigeration and are made at a zero waste to landfill facility.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (imperial)InteriorRequisitos eléctricos
TSG3005GD29,2 pies cúb.Blanco (pintado)208/230 V, 50 Hz
TSG1205GA12 pies cúb.Blanco (pintado)115 V/60 Hz
TSG1205GD12 pies cúb.Blanco (pintado)208-230 V/60 Hz
TSG2305GA23 pies cúb.Blanco (pintado)115 V/60 Hz
TSG2305GD23 pies cúb.Blanco (pintado)208-230 V/60 Hz
TSG3005GA29,2 pies cúb.Blanco (pintado)115 V/60 Hz
TSG4505GA45,8 pies cúb.Blanco (pintado)115 V/60 Hz
TSG4505GD45,8 pies cúb.Blanco (pintado)208-230 V/60 Hz
TSG5005GA51,1 pies cúb.Blanco (pintado)115 V/60 Hz
TSG5005GD51,1 pies cúb.Blanco (pintado)208-230 V/60 Hz
Número de catálogo TSG3005GD
Precio (MXN)
-
Capacidad (imperial):
29,2 pies cúb.
Interior:
Blanco (pintado)
Requisitos eléctricos:
208/230 V, 50 Hz
Thermo Scientific TSG Series Glass Door Laboratory Refrigerators offer quiet, intuitive operation with excellent temperature performance and an outstanding warranty, meeting the needs of a wide variety of professional users who need consistent, reliable performance without sacrificing usability or sustainability. All TSG Series Laboratory Refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigerants and are configured to provide excellent temperature performance while minimizing power consumption, all delivered from our zero waste to landfill facility in Asheville, North Carolina.

The drive for sample integrity and security

  • Designed to safely store media, reagents, and other laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned glass doors with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Extra and Alternate wire baskets, shelves, and drawers
  • Stainless Steel interior finish available
  • E-Lock Adapter kits
Especificaciones
Seguridad de accesoKey Lock
Capacidad del estante (imperial)100 lb.
Capacidad del estante (métrico)45 kg
Tipo de controlCapacitive Touch
DescripciónTSG Series General Purpose Refrigerator
Dimensiones (F x An x Al) exteriores37.3 x 34.0 x 78.5 in. (947 x 864 x 1994 mm)
Dimensiones interiores (F x An x Al)28.5 x 30.0 x 58.0 in. (723 x 762 x 1473 mm)
Requisitos eléctricos208/230 V, 50 Hz
Frecuencia60 Hz
InteriorBlanco (pintado)
Puertos1 Rear Access Port
RefrigeranteR290
Peso con embalaje (imperial)458 lb.
Peso con embalaje (métrico)208 kg
Temperatura (valor de referencia)+5°C
Garantía4 Years Parts and Labor + 1 Year Compressor
Material de la cabinaPainted Steel
Capacidad (imperial)29,2 pies cúb.
Capacidad (métrico)827 L
Certificaciones/conformidadcULus
Modelo de puertaVidrio
Acabado exteriorPintado
Opciones de vigilanciaSmartVue Pro, Remote Alarm Contacts
No. of Doors1
tipo de tapónNEMA 6-15
Estantes4 Ajustable
Intervalo de temperatura3°C a 7°C
Unit SizeEach