Thermo Scientific TSG Series Pharmacy Refrigerator
Thermo Scientific TSG Series Pharmacy Refrigerator
Thermo Scientific TSG Series Pharmacy Refrigerator
Thermo Scientific TSG Series Pharmacy Refrigerator
Thermo Scientific™

Thermo Scientific TSG Series Pharmacy Refrigerator

Green features
The TSG Pharmacy Refrigerators are purpose built for clinical and pharmaceutical applications needing 2-8°C, are NSF 456 certified and utilize SNAP and F-Gas compliant refrigeration. They are available in single and double door configurations and come with a 4-year parts and labor warranty.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(영국식 단위)인증/규정준수전기 요구사항
TSG3005PA29.2 cu. ft.cULus115V/60Hz
TSG1205PA12 cu. ft.cULus115 V/60Hz
TSG1205PD12 cu. ft.cULus208-230V/60Hz
TSG1205PV12 cu. ft.CE230V/50Hz
TSG2305PA23 cu. ft.cULus115V/60Hz
TSG2305PD23 cu. ft.cULus208-230V/60Hz
TSG2305PV23 cu. ft.CE230V/50Hz
TSG3005PD29.2 cu. ft.cULus208-230V/60Hz
TSG3005PV29.2 cu. ft.CE230V/50Hz
TSG5005PA51.1 cu. ft.cULus, ENERGYSTAR115V/60Hz
TSG5005PD51.1 cu. ft.cULus208-230V/60Hz
TSG5005PV51.1 cu. ft.CE230V/50Hz
카탈로그 번호 TSG3005PA
제품 가격(KRW)
-
용량(영국식 단위):
29.2 cu. ft.
인증/규정준수:
cULus
전기 요구사항:
115V/60Hz
Thermo Scientific TSG Series Pharmacy Refrigerators are specialized for clinical and pharmaceutical applications, offering quiet, intuitive operation with excellent temperature performance and an outstanding warranty. Validated NSF 456 compliant performance means TSG Series Pharmacy Refrigerators deliver exceptional security and temperature stability for vaccines, pharmaceuticals, and other precious cargo. All TSG Series Refrigerators are built with sustainability in mind to minimize power consumption without sacrificing performance at our zero waste to landfill facility in Asheville, NC.

Designed for sample integrity and security

  • Designed to safely store pharmaceuticals & vaccines requiring a temperature range of 2°C to 8°C
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and Certified to NSF/ANSI 456 Vaccine Storage standard and supports the US CDC, WHO, and other global vaccine storage guidelines and recommendations
  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle
  • Convenient, fully extendable and adjustable basket drawers
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Self-closing door with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Extra and alternate wire baskets, shelves, and drawers
  • E-Lock adapter kits
  • 4-20 milliamp transmitters and other remote alarm modules

Services and support

  • Industry Leading 60-month compressor warranty (varies by country, please contact us for details)
  • Included Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
  • Optional Validation Testing Programs include in field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs

 

사양
액세스 보안Key Lock
캐비닛 재질Painted Steel
용량(미터법) 선반27 kg
제어 유형Capacitive Touch
설명TSG Series Pharmacy Refrigerator
치수(깊이 x 폭 x 높이) 외부37.3 x 34.0 x 78.5 in. (947 x 864 x 1994 mm)
치수(깊이 x 폭 x 높이) 내부28.5 x 30.0 x 58.0 in. (723 x 762 x 1473 mm)
드로어Basket Drawers
전기 요구사항115V/60Hz
외부 마감재Painted
그린 기능ENERGY STAR
내부White (Painted)
모니터링 옵션SmartVue Pro, Remote Alarm Contacts
드로어 수6
포트1 Rear Access Port
냉매R290
배송 중량(영국식 단위)541 lb.
배송 중량(미터법)245 kg
온도(설정치)+5°C
워런티4 Years Parts and Labor + 1 Year Compressor
용량(영국식 단위)29.2 cu. ft.
용량(미터법)827 L
인증/규정준수cULus
도어 스타일Glass
No. of Doors2
플러그 유형NEMA 5-15
온도 범위3°C to 7°C
Unit SizeEach