TSG General Purpose Refrigerators - Boxed
Thermo Scientific™

TSG General Purpose Refrigerators - Boxed

The TSG Series of general purpose refrigerators are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
Have Questions?
變更視圖buttonViewtableView
產品號碼Capacity (Metric)Door StylePlug Type
TSG700REGBV700 LGlassEU
TSG1500REGBV1500 LGlassEU
TSG1500REGBW1500 LGlassUK
TSG1500RESBV1500 LSolidEU
TSG1500RESBW1500 LSolidUK
TSG400REGBV400 LGlassEU
TSG400REGBW400 LGlassUK
TSG400RESBV400 LSolidEU
TSG400RESBW400 LSolidUK
TSG700REGBW700 LGlassUK
TSG700RESBV700 LSolidEU
TSG700RESBW700 LSolidUK
產品號碼 TSG700REGBV
價格 (TWD)
-
Capacity (Metric):
700 L
Door Style:
Glass
Plug Type:
EU
The Thermo Scientific™ TSG Series general purpose laboratory refrigerators are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series General Purpose Refrigerators are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from 400 L to 1500 L double door
  • Glass door (triple pane) and solid door configurations
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
規格
Access SecurityKey Lock
Cabinet MaterialSteel
Capacity (Metric) Shelf30 kg
CastersYes
Control TypeDigital Microprocessor
DefrostAdaptive Defrost
DescriptionTSG Series General Purpose Laboratory Refrigerator
Dimensions (D x W x H) Interior686 x 600 x 1505 mm
Dimensions (L x W x H) Exterior855 x 750 x 2085 mm
Electrical Requirements230 V/50 Hz
Exterior FinishPainted Steel
Frequency50 Hz
InteriorPainted Steel
Monitoring OptionsAlarm
Packaging TypeBoxed
Ports1 Rear Access Port
RefrigerantR290
Shelves4 Adjustable
Shipping Weight (Metric)175 kg
Smart View CompatibilitySmart-Vue Pro Compatible
Temperature (Setpoint)+5°C
Warranty36 Months
Capacity (Metric)700 L
Certifications/ComplianceCE
Door StyleGlass
No. of Doors1
Plug TypeEU
Temperature Range2°C to 10°C
Voltage230 V
Unit SizeEach