Search
| Catalog Number | Type | ACT Label |
|---|---|---|
| TSQ03-11001 | TSQ LC-MS with Dry Pump | Yes |
| TSQ03-10001 | TSQ LC-MS with Oil Pump | No |
| B51006570 | TSQ LC-MS with Oil Pump | No |
Achieve next-level productivity, versatility, and performance for method development and routine sample analysis. The TSQ Quantis Plus triple quadrupole mass spectrometer combines enhanced acquisition speed and sensitivity with proven robustness and reliability to deliver confident targeted screening and precise quantitation for high-throughput applications. Advanced instrument control software, compound database integration, and predefined method templates simplify operation, enabling LC-MS/MS users of all skill levels to obtain high-confidence results.
The TSQ Quantis Plus mass spectrometer seamlessly integrates with industry-leading quantitation software, (U)HPLC and HPIC systems, and sample preparation solutions; it also offers enhanced efficiency for targeted screening and quantitation workflows for a wide array of molecular compounds. Other benefits include:
Thermo Fisher Scientific is working to provide more information about our products' environmental impact by stepping up to participate in the ACT Label program with selected products.
Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.
For General Lab Use Only. Not For Diagnostic Procedures.