TSQ Fortis™ Plus Triple Quadrupole Mass Spectrometer
TSQ Fortis™ Plus Triple Quadrupole Mass Spectrometer
TSQ Fortis™ Plus Triple Quadrupole Mass Spectrometer
TSQ Fortis™ Plus Triple Quadrupole Mass Spectrometer
Thermo Scientific™
TSQ Fortis™ Plus Triple Quadrupole Mass Spectrometer
Achieve quantitative productivity for high-throughput workflows with enhanced acquisition speeds, sensitivity, and operational simplicity.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoMass RangeType
TSQ03-110032 to 3000 m/zTSQ LC-MS with Dry Pump
TSQ03-100032 to 3000 m/zTSQ LC-MS with Oil Pump
Número do catálogo TSQ03-11003
Preço (BRL)
-
Mass Range:
2 to 3000 m/z
Type:
TSQ LC-MS with Dry Pump
Accelerate your path to quantitative productivity with the Thermo Scientific™ TSQ Fortis™ Plus triple quadrupole mass spectrometer. Enhanced acquisition speed and sensitivity, combined with proven robustness and reliability, deliver next-level performance and value for high-throughput analyses. Simplified operations integrated with powerful Thermo Scientific™ chromatographic systems and market-leading software ensure that users of all skill levels obtain reproducible, high-confidence results to meet laboratory requirements.
The TSQ Fortis Plus mass spectrometer seamlessly integrates with industry-leading quantitative software, (U)HPLC and HPIC systems, and sample preparation solutions. It also offers enhanced productivity for targeted quantitation workflows with a wide array of molecular compounds. Other benefits include:
  • Increased SRM acquisition speeds providing greater sample coverage per analysis and higher throughput with excellent quantitative performance
  • Direct method transfer from the TSQ Fortis mass spectrometer enabling laboratory scalability
  • Advanced instrument controls and easy calibration procedures simplifying operations
  • Outstanding robustness delivering enhanced confidence in data acquisition
Especificações
For Use With (Application)Pharmaceutical, Environmental, Food Safety, Clinical Research, Forensic Toxicology
For Use With (Equipment)OptaMax NG ion source, VeriSpray PaperSpray ion source
Mass Range2 to 3000 m/z
TypeTSQ LC-MS with Dry Pump
Unit SizeEach