Thermo Scientific™

TSV Value Undercounter Refrigerator

Catalog number: TSV05RPAA
Thermo Scientific™

TSV Value Undercounter Refrigerator

Catalog number: TSV05RPAA
Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Undercounter Refrigerator
 
Catalog Number
TSV05RPAA
Unit Size
Each
Type
General Purpose ADA Refrigerator
Price (USD)
Full specifications
Temperature Range1°C to 12°C
Certifications/CompliancecULus
Capacity (English)5.5 cu. ft.
Capacity (Metric)155.7 L
Electrical Requirements115V 60Hz
Voltage115 V
Frequency60Hz
Amperage.8A
Plug TypeNEMA 5-15 P
Dimensions (D x W x H) Interior17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5 cm)
Shipping Weight (English)95 lbs.
Shipping Weight (Metric)43 kg
RefrigerantR600a hydrocarbon refrigerant
InteriorABS Plastic
Access SecurityKey Lock
Warranty12 Months
ColorWhite
Exterior FinishEnameled Steel
Energy Usage.31 kWh/Day
Regulatory ApprovalsADA Compliant
For Use With (Application)Indoor General Storage of Non-Critical Samples and Reagents
Typical Lifetime10 Years
Min. Clearance4 in. on all sides
Temperature (Setpoint)Adjustable
Casters2 adjustable feet in front
DefrostManual
Dimensions (DxWxH) Exterior23.5 x 23.63 x 33.5 in. (59.69 x 60.02 x 85.1cm)
TypeGeneral Purpose ADA Refrigerator
Control TypeMechanical Dial
Door StyleHinged, Open Left to Right
CompressorHermetic Compressor
InsulationCyclopentane
No. of Doors1
Shelves3 Adjustable
Unit SizeEach
Catalog Number
Unit Size
Each
Type
General Purpose Refrigerator
Price (USD)
Full specifications
Temperature Range1°C to 12°C
Certifications/CompliancecULus
Capacity (English)5.5 cu. ft.
Capacity (Metric)155.7 L
Electrical Requirements115V 60Hz
Voltage115 V
Frequency60Hz
Amperage.8A
Plug TypeNEMA 5-15 P
Dimensions (D x W x H) Interior17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm)
Shipping Weight (English)95 lbs.
Shipping Weight (Metric)43 kg
RefrigerantR600a hydrocarbon refrigerant
InteriorABS Plastic
Access SecurityKey Lock
Warranty12 months
ColorWhite
Exterior FinishEnameled Steel
Energy Usage.31 kWh/Day
For Use With (Application)Indoor General Storage of Non-Critical Samples and Reagents
Typical Lifetime10 years
Min. Clearance4 in. on all sides
Temperature (Setpoint)Adjustable
Casters2 Adjustable Feet in Front
DefrostManual
Dimensions (DxWxH) Exterior23.5 x 23.63 x 33.5 in. (59.69 x 60.02 x 85.1cm)
TypeGeneral Purpose Refrigerator
Control TypeMechanical Dial
Door StyleHinged, open left to right
CompressorHermetic Compressor
InsulationCyclopentane
No. of Doors1
Shelves3 adjustable
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeTypePrice (USD)
TSV05RPAAFull specifications
EachGeneral Purpose ADA RefrigeratorRequest A Quote
Temperature Range1°C to 12°C
Certifications/CompliancecULus
Capacity (English)5.5 cu. ft.
Capacity (Metric)155.7 L
Electrical Requirements115V 60Hz
Voltage115 V
Frequency60Hz
Amperage.8A
Plug TypeNEMA 5-15 P
Dimensions (D x W x H) Interior17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5 cm)
Shipping Weight (English)95 lbs.
Shipping Weight (Metric)43 kg
RefrigerantR600a hydrocarbon refrigerant
InteriorABS Plastic
Access SecurityKey Lock
Warranty12 Months
ColorWhite
Exterior FinishEnameled Steel
Energy Usage.31 kWh/Day
Regulatory ApprovalsADA Compliant
For Use With (Application)Indoor General Storage of Non-Critical Samples and Reagents
Typical Lifetime10 Years
Min. Clearance4 in. on all sides
Temperature (Setpoint)Adjustable
Casters2 adjustable feet in front
DefrostManual
Dimensions (DxWxH) Exterior23.5 x 23.63 x 33.5 in. (59.69 x 60.02 x 85.1cm)
TypeGeneral Purpose ADA Refrigerator
Control TypeMechanical Dial
Door StyleHinged, Open Left to Right
CompressorHermetic Compressor
InsulationCyclopentane
No. of Doors1
Shelves3 Adjustable
Unit SizeEach
TSV05RPSAFull specifications
EachGeneral Purpose RefrigeratorRequest A Quote
Temperature Range1°C to 12°C
Certifications/CompliancecULus
Capacity (English)5.5 cu. ft.
Capacity (Metric)155.7 L
Electrical Requirements115V 60Hz
Voltage115 V
Frequency60Hz
Amperage.8A
Plug TypeNEMA 5-15 P
Dimensions (D x W x H) Interior17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm)
Shipping Weight (English)95 lbs.
Shipping Weight (Metric)43 kg
RefrigerantR600a hydrocarbon refrigerant
InteriorABS Plastic
Access SecurityKey Lock
Warranty12 months
ColorWhite
Exterior FinishEnameled Steel
Energy Usage.31 kWh/Day
For Use With (Application)Indoor General Storage of Non-Critical Samples and Reagents
Typical Lifetime10 years
Min. Clearance4 in. on all sides
Temperature (Setpoint)Adjustable
Casters2 Adjustable Feet in Front
DefrostManual
Dimensions (DxWxH) Exterior23.5 x 23.63 x 33.5 in. (59.69 x 60.02 x 85.1cm)
TypeGeneral Purpose Refrigerator
Control TypeMechanical Dial
Door StyleHinged, open left to right
CompressorHermetic Compressor
InsulationCyclopentane
No. of Doors1
Shelves3 adjustable
Unit SizeEach
Showing 2 of 2
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish

Figures