Specifications
Access SecurityKey Lock
Amperage0.8 A
Casters2 Adjustable Feet in Front
ColorWhite
Control TypeMechanical Dial
DefrostManual
Dimensions (DxWxH) Exterior23.5 x 23.63 x 33.5 in. (59.69 x 60.02 x 85.1cm)
Dimensions (D x W x H) Interior17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm)
Electrical Requirements115V 60Hz
Energy Usage.31 kWh/Day
For Use With (Application)Indoor General Storage of Non-Critical Samples and Reagents
Frequency60 Hz
Hinge SideRight
InsulationCyclopentane
InteriorABS Plastic
Min. Clearance4 in. on all sides
RefrigerantR600a hydrocarbon refrigerant
Shipping Weight (English)95 lb.
Shipping Weight (Metric)43 kg
Temperature (Setpoint)Adjustable
TypeGeneral Purpose Refrigerator
Typical Lifetime10 years
Warranty12 months
Capacity (English)5.5 cu. ft.
Capacity (Metric)155.7 L
Certifications/CompliancecULus
CompressorHermetic
Door StyleHinged
Exterior FinishEnameled Steel
No. of Doors1
Plug TypeNEMA 5-15 P
Shelves3 Adjustable
Temperature Range1°C to 12°C
Voltage115 V
Unit SizeEach