TSV Value Convertible Freezer/Refrigerator
TSV Value Convertible Freezer/Refrigerator
TSV Value Convertible Freezer/Refrigerator
TSV Value Convertible Freezer/Refrigerator
TSV Value Convertible Freezer/Refrigerator
TSV Value Convertible Freezer/Refrigerator
TSV Value Convertible Freezer/Refrigerator
TSV Value Convertible Freezer/Refrigerator
TSV Value Convertible Freezer/Refrigerator
Thermo Scientific™

TSV Value Convertible Freezer/Refrigerator

Flexible and economical laboratory storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
TSV20FPSAGeneral Purpose Freezer (Convertible to Refrigerator)
TSV20RPSAGeneral Purpose Refrigerator (Convertible to Freezer)
Catalog number TSV20FPSA
Price (MXN)
-
Type:
General Purpose Freezer (Convertible to Refrigerator)
Thermo Scientific TSV Value freezers and refrigerators offer economical and simple sample storage while also offering increased flexibility of operating as either a refrigerator or a freezer with the push of a button.

The TSV20FSPSA ships as an auto defrost freezer and the TSV20RPSA ships as a refrigerator, but each easily converts to a refrigerator or an auto defrost freezer respectively.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clean, durable finish
Specifications
Access SecurityNone
Amperage2.2A
Cabinet MaterialCold-rolled Steel
Casters2 adjustable leveling feet
ColorWhite
Control TypeElectronic control
DefrostAutomatic
Dimensions (D x W x H) Interior22.5 x 26.5 x 69 in. (57.15 x 67.3 x 175.26 cm)
Dimensions (L x W x H) Exterior28.94 x 32.76 x 76.77 in. (73.5 x 83.2 x 195 cm)
Door Shelves4 fixed shelves
Electrical Requirements115 V/60 Hz
Exterior FinishWhite-enameled Steel
For Use With (Application)Indoor general storage of non-critical samples and reagents
Frequency50 Hz
Hinge SideRight
InsulationCyclopentane
InteriorABS plastic
LightingInterior - Yes
Min. Clearance4 in. on sides, 4 in. at back, 11 in. on top
Monitoring OptionsRemote Alarm Contacts
Operating Modes2 - refrigerator or freezer
RefrigerantR600a
Shelves3 Adjustable
Shipping Weight (English)244 lb.
Shipping Weight (Metric)110.6 kg
Temperature (Setpoint)Adjustable
TypeGeneral Purpose Freezer (Convertible to Refrigerator)
Typical Lifetime10 Years
Warranty13 Months Parts and Labor
Capacity (English)21 cu. ft.
Capacity (Metric)594 L
Certifications/CompliancecULus, cULus, cULus, cULus, cULus
CompressorBottom-mounted
Door StyleHinged
No. of Doors1 (1 compartment)
Plug TypeNEMA 5-15 P
StyleUpright
Temperature Range1°C to 12°C or -12°C to -24°C
Voltage115 V
Unit SizeEach