TSV Series 72 cu.ft. Lab Refrigerator
Thermo Scientific™

TSV Series 72 cu.ft. Lab Refrigerator

Green features
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents. Featuring three self-closing glass doors, the 72 cu. ft. TSV Series Lab Refrigerators offer an economical solution for users who need significant space for temperature controlled storage.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoMaterial de la cabinaAcabado exterior
TSV72RPGAAcero pintadoAcero pintado
TSV72RSGAAcero inoxidableAcero inoxidable
Número de catálogo TSV72RPGA
Precio (MXN)
-
Material de la cabina:
Acero pintado
Acabado exterior:
Acero pintado
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents. Featuring three self-closing glass doors, the 72 cu. ft. TSV Series Lab Refrigerators offer an economical solution for users who need significant space for temperature controlled storage. Additionally, TSV Series 72 cu.ft. Lab Refrigerators come with remote alarm contacts, high and low temperature alarms, and an adjustbale setpoint with a digital controller.
  • Three self-closing glass doors
  • Remote alarm contacts
  • High and low temperature alarms
  • Digital controller with adjustable temperature setpoint
  • Door ajar alarm
  • Optional stainless steel interior
Especificaciones
Seguridad de accesoKey lock
Material de la cabinaAcero pintado
Capacidad del estante (imperial)100 lb.
Capacidad del estante (métrico)45 kg
Ruedas giratoriasYes, customer installed
DescongelaciónAutomático
Dimensiones (F x An x Al) exteriores75.7 x 32 x 81 in. (1924 x 813 x 2057 mm)
Dimensiones interiores (F x An x Al)72 x 28.5 x 61.75 in. (1829 x 724 x 1569 mm)
PantallaDigital
Requisitos eléctricos120 V/60Hz
Acabado exteriorAcero pintado
Características ecológicasENERGY STAR
Opciones de vigilanciaContactos de alarma remota compatibles con SMART VUE
PuertosOne rear access port
RefrigeranteR290
Estantes12 Adjustable
Peso con embalaje (imperial)730 lb.
Peso con embalaje (métrico)318 kg
Temperatura (valor de referencia)5°C
Garantía2 years parts & labor, 3 additional years compressor parts
Capacidad (imperial)72 cu. ft.
Capacidad (métrico)2038 L
Certificaciones/conformidadcULus
compresorBottom-mounted
Modelo de puertaGlass Self-closing with Lock
No. of Doors3
tipo de tapónNEMA 5-15
ModeloIndependiente
Intervalo de temperatura4°C to 6°C
Unit SizeEach