Proteome Discoverer™ Software eLearning Course
Thermo Scientific™

Proteome Discoverer™ Software eLearning Course

Proteome Discoverer™ Basic operations eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Catalog NumberDuration
TSWPDISBOP4 to 8 hr.
Catalog number TSWPDISBOP
Price (USD)
310.00
Each
Duration:
4 to 8 hr.
Price (USD)
310.00
Each
Proteome Discoverer™ Basic operations is an introduction to the basic operations of Proteome Discoverer™. It will teach the fundamentals of setting up a basic protein identification search using the Proteome Discoverer™ 2.1 software. This course contains a software overview, how to set up a study, parameters needed for a processing and consensus workflow, and opening and viewing results.

The Proteome Discoverer™ Basic Operations eLearning course is an introduction to the basic training of the Proteome Discoverer™

software. Topics and concepts discussed include:

  • Proteome Discoverer™ Basic ID Search
  • Proteome Discoverer™ TMT Analysis Workflow
  • Proteome Discoverer™ and Software Label-free Quantitation Demonstration
Specifications
Access Availablity30 days upon end user first acess
Course TopicBasic Operations
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelRoutine
Duration4 to 8 hr.
For Use With (Equipment)Proteome Discoverer
Product LineThermo Scientific
Product TypeSoftware
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach