TraceFinder™ Software eLearning Course
Thermo Scientific™

TraceFinder™ Software eLearning Course

The TraceFinder™ Basic Operations Basic operations eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Catalog NumberDuration
TSWTFBOP4 to 8 hr.
Catalog number TSWTFBOP
Price (MXN)
-
Duration:
4 to 8 hr.
The TraceFinder™ Basic Operations eLearning course is an introduction to the basic principles of the TraceFinder™ software. It will teach the fundamentals of general quantitation routine analysis workflow, general quantitation method development workflow, and HRAM targeted screening.

The TraceFinder™ Basic Operations eLearning courseis an introduction to the TraceFinder software. Topics and concepts discussed include:

  • TraceFinder™ General Quantitation Routine Analysis Workflow
  • TraceFinder™ Software General Quantitation Method Development Workflow
  • TraceFinder™ HRAM Targeted Screening

The TraceFinder™ general quantitation routine analysis workflow module will help users get familiar with some of the basic concepts in the TraceFinder™ software. It will demonstrate the use of TraceFinder™ software for data acquisition and analysis with an existing analysis method (Master Method), basic operations of creating a batch, submitting it for acquisition, monitoring the analysis, reviewing the data, and generating reports.

The TraceFinder™ software general quantitation method development workflow module will help users get familiar with some of the basic concepts in the method development mode of the TraceFinder™ Software. It will demonstrate the use of TraceFinder™ software for working with compound databases and creating Master Methods for the quantitation workflow.

TraceFinder™ HRAM targeted screening module provides training on targeted screening method building for HRAM data.

Specifications
Access Availablity30 days upon end user first acess
Course TopicBasic Operations
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelRoutine
Duration4 to 8 hr.
For Use With (Equipment)TraceFinder
Product LineThermo Scientific
Product TypeSoftware
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach