Thermo Scientific™

TSX Series High-Performance Pharmacy Refrigerators

Green features
Catalog number: TSX3005PD
Thermo Scientific™

TSX Series High-Performance Pharmacy Refrigerators

Green features
Catalog number: TSX3005PD
Thermo Scientific™ TSX Series high-performance pharmacy refrigerators are designed with features that support sample protection and sustainability objectives for the storage of pharmaceuticals, vaccines, chemotherapy and other medical and pharmacy-grade storage requiring 2° to 8°C. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection. Now certified to NSF/ANSI 456 Vaccine Storage standard, the TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
 
Catalog Number
TSX3005PD
Unit Size
Each
Plug Type
NEMA 6-15
Electrical Requirements
208-230V, 60Hz
Dimensions (D x W x H) Interior
28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Door Style
Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Electrical Requirements208-230V, 60Hz
Voltage208/230 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 6-15
Dimensions (L x W x H) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Weight (English)473 lb.
Weight (Metric)214 kg
Shipping Weight (English)541 lb.
Shipping Weight (Metric)245 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers6
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.3 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior30 in.
Width (Metric) Interior76.2 cm
DefrostAutomatic
Width (English) Exterior34.0 in.
Width (Metric) Exterior86.4 cm
Dimensions (DxWxH) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Depth (English) Exterior37.3 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior94.7 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 5-15
Electrical Requirements
115V, 60Hz
Dimensions (D x W x H) Interior
28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
Door Style
Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)23 cu. ft.
Capacity (Metric)650 L
Electrical Requirements115V, 60Hz
Voltage115 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 5-15
Dimensions (L x W x H) Exterior37.8 x 28 x 78.6 in. (96.2 x 71.1 x 199.6 cm)
Dimensions (D x W x H) Interior28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
Weight (English)446 lb.
Weight (Metric)211 kg
Shipping Weight (English)529 lb.
Shipping Weight (Metric)240 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers6
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.3 cm
DrawersBasket Drawers
Height (English) Exterior78.6 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.6 cm
Height (Metric) Interior147.3 cm
Width (English) Interior24 in.
Width (Metric) Interior61 cm
DefrostAutomatic
Width (English) Exterior28 in.
Width (Metric) Exterior71.1 cm
Depth (English) Exterior37 in.
TypePharmacy Refrigerator
Depth (Metric) Exterior96.2 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 5-15
Electrical Requirements
115V, 60Hz
Dimensions (D x W x H) Interior
28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Door Style
Double Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR, NSF 456
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Electrical Requirements115V, 60Hz
Voltage115 V
Frequency60 Hz
Amperage15 A (CB)
Plug TypeNEMA 5-15
Dimensions (L x W x H) Exterior37.9 x 56.5 x 78.5 in. (96.2 x 143.5 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Weight (English)744
Weight (Metric)337
Shipping Weight (English)830
Shipping Weight (Metric)376
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
WarrantyWarranty varies by country, please contact us for details
No. of Drawers8
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.4 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior52.5 in.
Width (Metric) Interior133.4 cm
DefrostAutomatic
Width (English) Exterior56.5 in.
Width (Metric) Exterior143.5 cm
Depth (English) Exterior37.8 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior96.0 cm
Green FeaturesEnergy efficient
Door StyleDouble Glass
Monitoring OptionsAlarm, Recorder
No. of Doors2
Shelves8
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 6-15
Electrical Requirements
208-230V, 60Hz
Dimensions (D x W x H) Interior
28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Door Style
Double Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR, NSF 456
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Electrical Requirements208-230V, 60Hz
Voltage208/230 V
Frequency50/60 Hz
Amperage15 A (CB)
Plug TypeNEMA 6-15
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Weight (English)744
Weight (Metric)337
Shipping Weight (English)830
Shipping Weight (Metric)376
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
WarrantyWarranty varies by country, please contact us for details
No. of Drawers8
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.4 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior52.5 in.
Width (Metric) Interior133.4 cm
DefrostAutomatic
Width (English) Exterior56.5 in.
Width (Metric) Exterior143.5 cm
Depth (English) Exterior37.8 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior96.0 cm
Green FeaturesEnergy efficient
Door StyleDouble Glass
Monitoring OptionsAlarm, Recorder
No. of Doors2
Shelves8
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
CEE 7/7
Electrical Requirements
208-230V, 50Hz
Dimensions (D x W x H) Interior
28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Door Style
Double Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, CE
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Electrical Requirements208-230V, 50Hz
Voltage208/230 V
Frequency50/60 Hz
Amperage15 A (CB)
Plug TypeCEE 7/7
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Weight (English)744
Weight (Metric)337
Shipping Weight (English)830
Shipping Weight (Metric)376
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
WarrantyWarranty varies by country, please contact us for details
No. of Drawers8
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.4 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior52.5 in.
Width (Metric) Interior133.4 cm
DefrostAutomatic
Width (English) Exterior56.5 in.
Width (Metric) Exterior143.5 cm
Depth (English) Exterior37.8 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior96.0 cm
Green FeaturesEnergy efficient
Door StyleDouble Glass
Monitoring OptionsAlarm, Recorder
No. of Doors2
Rack Capacity12
Shelves8
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 5-15
Electrical Requirements
115V, 60Hz
Dimensions (D x W x H) Interior
21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Door Style
Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Electrical Requirements115V, 60Hz
Voltage115 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 5-15
Dimensions (L x W x H) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Weight (English)334 lb.
Weight (Metric)151 kg
Shipping Weight (English)388 lb.
Shipping Weight (Metric)176 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers5
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior21.8 in.
Depth (Metric) Interior55.4 cm
DrawersBasket Drawers
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Interior20 in.
Width (Metric) Interior50.8 cm
DefrostAutomatic
Width (English) Exterior24.0 in.
Width (Metric) Exterior61.9 cm
Dimensions (DxWxH) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Depth (English) Exterior30.5 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior77.5 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 6-15
Electrical Requirements
208-230V, 60Hz
Dimensions (D x W x H) Interior
21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Door Style
Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Electrical Requirements208-230V, 60Hz
Voltage208/230 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 6-15
Dimensions (L x W x H) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Weight (English)334 lb.
Weight (Metric)151 kg
Shipping Weight (English)388 lb.
Shipping Weight (Metric)176 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers5
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior21.8 in.
Depth (Metric) Interior55.4 cm
DrawersBasket Drawers
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Interior20 in.
Width (Metric) Interior50.8 cm
DefrostAutomatic
Width (English) Exterior24.0 in.
Width (Metric) Exterior61.9 cm
Dimensions (DxWxH) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Depth (English) Exterior30.5 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior77.5 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
CEE 7/7
Electrical Requirements
208-230V, 50Hz
Dimensions (D x W x H) Interior
21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Door Style
Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, CE
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Electrical Requirements208-230V, 50Hz
Voltage230 V
Frequency50 Hz
Amperage15 A
Plug TypeCEE 7/7
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Weight (English)334 lb.
Weight (Metric)151 kg
Shipping Weight (English)388 lb.
Shipping Weight (Metric)176 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers5
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior21.8 in.
Depth (Metric) Interior55.4 cm
DrawersBasket Drawers
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Interior20 in.
Width (Metric) Interior50.8 cm
DefrostAutomatic
Width (English) Exterior24.0 in.
Width (Metric) Exterior61.9 cm
Dimensions (DxWxH) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Depth (English) Exterior30.5 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior77.5 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Rack Capacity5
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 5-15
Electrical Requirements
115V, 60Hz
Dimensions (D x W x H) Interior
28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Door Style
Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Electrical Requirements115V, 60Hz
Voltage115 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 5-15
Dimensions (L x W x H) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Weight (English)473 lb.
Weight (Metric)214 kg
Shipping Weight (English)541 lb.
Shipping Weight (Metric)245 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers6
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.3 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior30 in.
Width (Metric) Interior76.2 cm
DefrostAutomatic
Width (English) Exterior34.0 in.
Width (Metric) Exterior86.4 cm
Dimensions (DxWxH) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Depth (English) Exterior37.3 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior94.7 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
CEE 7/7
Electrical Requirements
208-230V, 50Hz
Dimensions (D x W x H) Interior
28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Door Style
Glass
Price (USD)
Full specifications
Product LineTSX Series
Certifications/ComplianceUL, cUL, CE
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Electrical Requirements208-230V, 50Hz
Voltage230 V
Frequency50 Hz
Amperage15 A
Plug TypeCEE 7/7
Dimensions (L x W x H) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Weight (English)473 lb.
Weight (Metric)214 kg
Shipping Weight (English)541 lb.
Shipping Weight (Metric)245 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers6
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.3 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior30 in.
Width (Metric) Interior76.2 cm
DefrostAutomatic
Width (English) Exterior34.0 in.
Width (Metric) Exterior86.4 cm
Dimensions (DxWxH) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Depth (English) Exterior37.3 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior94.7 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Rack Capacity6
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizePlug TypeElectrical RequirementsDimensions (D x W x H) InteriorDoor StylePrice (USD)
TSX3005PDFull specifications
EachNEMA 6-15208-230V, 60Hz28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Electrical Requirements208-230V, 60Hz
Voltage208/230 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 6-15
Dimensions (L x W x H) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Weight (English)473 lb.
Weight (Metric)214 kg
Shipping Weight (English)541 lb.
Shipping Weight (Metric)245 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers6
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.3 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior30 in.
Width (Metric) Interior76.2 cm
DefrostAutomatic
Width (English) Exterior34.0 in.
Width (Metric) Exterior86.4 cm
Dimensions (DxWxH) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Depth (English) Exterior37.3 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior94.7 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
TSX2305PAFull specifications
EachNEMA 5-15115V, 60Hz28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)23 cu. ft.
Capacity (Metric)650 L
Electrical Requirements115V, 60Hz
Voltage115 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 5-15
Dimensions (L x W x H) Exterior37.8 x 28 x 78.6 in. (96.2 x 71.1 x 199.6 cm)
Dimensions (D x W x H) Interior28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
Weight (English)446 lb.
Weight (Metric)211 kg
Shipping Weight (English)529 lb.
Shipping Weight (Metric)240 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers6
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.3 cm
DrawersBasket Drawers
Height (English) Exterior78.6 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.6 cm
Height (Metric) Interior147.3 cm
Width (English) Interior24 in.
Width (Metric) Interior61 cm
DefrostAutomatic
Width (English) Exterior28 in.
Width (Metric) Exterior71.1 cm
Depth (English) Exterior37 in.
TypePharmacy Refrigerator
Depth (Metric) Exterior96.2 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
TSX5005PAFull specifications
EachNEMA 5-15115V, 60Hz28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)Double GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR, NSF 456
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Electrical Requirements115V, 60Hz
Voltage115 V
Frequency60 Hz
Amperage15 A (CB)
Plug TypeNEMA 5-15
Dimensions (L x W x H) Exterior37.9 x 56.5 x 78.5 in. (96.2 x 143.5 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Weight (English)744
Weight (Metric)337
Shipping Weight (English)830
Shipping Weight (Metric)376
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
WarrantyWarranty varies by country, please contact us for details
No. of Drawers8
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.4 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior52.5 in.
Width (Metric) Interior133.4 cm
DefrostAutomatic
Width (English) Exterior56.5 in.
Width (Metric) Exterior143.5 cm
Depth (English) Exterior37.8 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior96.0 cm
Green FeaturesEnergy efficient
Door StyleDouble Glass
Monitoring OptionsAlarm, Recorder
No. of Doors2
Shelves8
Unit SizeEach
TSX5005PDFull specifications
EachNEMA 6-15208-230V, 60Hz28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)Double GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR, NSF 456
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Electrical Requirements208-230V, 60Hz
Voltage208/230 V
Frequency50/60 Hz
Amperage15 A (CB)
Plug TypeNEMA 6-15
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Weight (English)744
Weight (Metric)337
Shipping Weight (English)830
Shipping Weight (Metric)376
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
WarrantyWarranty varies by country, please contact us for details
No. of Drawers8
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.4 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior52.5 in.
Width (Metric) Interior133.4 cm
DefrostAutomatic
Width (English) Exterior56.5 in.
Width (Metric) Exterior143.5 cm
Depth (English) Exterior37.8 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior96.0 cm
Green FeaturesEnergy efficient
Door StyleDouble Glass
Monitoring OptionsAlarm, Recorder
No. of Doors2
Shelves8
Unit SizeEach
TSX5005PVFull specifications
EachCEE 7/7208-230V, 50Hz28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)Double GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, CE
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Electrical Requirements208-230V, 50Hz
Voltage208/230 V
Frequency50/60 Hz
Amperage15 A (CB)
Plug TypeCEE 7/7
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Weight (English)744
Weight (Metric)337
Shipping Weight (English)830
Shipping Weight (Metric)376
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
WarrantyWarranty varies by country, please contact us for details
No. of Drawers8
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.4 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior52.5 in.
Width (Metric) Interior133.4 cm
DefrostAutomatic
Width (English) Exterior56.5 in.
Width (Metric) Exterior143.5 cm
Depth (English) Exterior37.8 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior96.0 cm
Green FeaturesEnergy efficient
Door StyleDouble Glass
Monitoring OptionsAlarm, Recorder
No. of Doors2
Rack Capacity12
Shelves8
Unit SizeEach
TSX1205PAFull specifications
EachNEMA 5-15115V, 60Hz21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Electrical Requirements115V, 60Hz
Voltage115 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 5-15
Dimensions (L x W x H) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Weight (English)334 lb.
Weight (Metric)151 kg
Shipping Weight (English)388 lb.
Shipping Weight (Metric)176 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers5
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior21.8 in.
Depth (Metric) Interior55.4 cm
DrawersBasket Drawers
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Interior20 in.
Width (Metric) Interior50.8 cm
DefrostAutomatic
Width (English) Exterior24.0 in.
Width (Metric) Exterior61.9 cm
Dimensions (DxWxH) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Depth (English) Exterior30.5 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior77.5 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
TSX1205PDFull specifications
EachNEMA 6-15208-230V, 60Hz21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Electrical Requirements208-230V, 60Hz
Voltage208/230 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 6-15
Dimensions (L x W x H) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Weight (English)334 lb.
Weight (Metric)151 kg
Shipping Weight (English)388 lb.
Shipping Weight (Metric)176 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers5
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior21.8 in.
Depth (Metric) Interior55.4 cm
DrawersBasket Drawers
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Interior20 in.
Width (Metric) Interior50.8 cm
DefrostAutomatic
Width (English) Exterior24.0 in.
Width (Metric) Exterior61.9 cm
Dimensions (DxWxH) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Depth (English) Exterior30.5 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior77.5 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
TSX1205PVFull specifications
EachCEE 7/7208-230V, 50Hz21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, CE
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Electrical Requirements208-230V, 50Hz
Voltage230 V
Frequency50 Hz
Amperage15 A
Plug TypeCEE 7/7
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Weight (English)334 lb.
Weight (Metric)151 kg
Shipping Weight (English)388 lb.
Shipping Weight (Metric)176 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers5
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior21.8 in.
Depth (Metric) Interior55.4 cm
DrawersBasket Drawers
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Interior20 in.
Width (Metric) Interior50.8 cm
DefrostAutomatic
Width (English) Exterior24.0 in.
Width (Metric) Exterior61.9 cm
Dimensions (DxWxH) Exterior30.5 x 24.0 x 73.0 in. (77.5 x 61.9 x 185.4 cm)
Depth (English) Exterior30.5 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior77.5 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Rack Capacity5
Unit SizeEach
TSX3005PAFull specifications
EachNEMA 5-15115V, 60Hz28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, ENERGY STAR
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Electrical Requirements115V, 60Hz
Voltage115 V
Frequency60 Hz
Amperage15 A
Plug TypeNEMA 5-15
Dimensions (L x W x H) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Weight (English)473 lb.
Weight (Metric)214 kg
Shipping Weight (English)541 lb.
Shipping Weight (Metric)245 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers6
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.3 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior30 in.
Width (Metric) Interior76.2 cm
DefrostAutomatic
Width (English) Exterior34.0 in.
Width (Metric) Exterior86.4 cm
Dimensions (DxWxH) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Depth (English) Exterior37.3 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior94.7 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Unit SizeEach
TSX3005PVFull specifications
EachCEE 7/7208-230V, 50Hz28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)GlassRequest A Quote
Product LineTSX Series
Certifications/ComplianceUL, cUL, CE
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Electrical Requirements208-230V, 50Hz
Voltage230 V
Frequency50 Hz
Amperage15 A
Plug TypeCEE 7/7
Dimensions (L x W x H) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Weight (English)473 lb.
Weight (Metric)214 kg
Shipping Weight (English)541 lb.
Shipping Weight (Metric)245 kg
DisplayMicroprocessor
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Chart RecorderOptional
No. of Drawers6
Temperature (Setpoint)3°-7°C (5°C)
Depth (English) Interior28.5 in.
Depth (Metric) Interior72.3 cm
DrawersBasket Drawers
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Interior30 in.
Width (Metric) Interior76.2 cm
DefrostAutomatic
Width (English) Exterior34.0 in.
Width (Metric) Exterior86.4 cm
Dimensions (DxWxH) Exterior37.3 x 34.0 x 78.5 in. (94.7 x 86.4 x 199.4 cm)
Depth (English) Exterior37.3 in.
TypeHigh-Performance Refrigerator
Depth (Metric) Exterior94.7 cm
Green FeaturesEnergy efficient
Door StyleGlass
Monitoring OptionsAlarm, Recorder
No. of Doors1
Rack Capacity6
Unit SizeEach
Showing 10 of 12Show more
  • Designed to safely store pharmaceuticals,  vaccines, chemotherapy and other medical-grade storage requiring a temperature range of 2°C to 8°C
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Up to 40% less energy usage than conventional refrigerant models*
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Operating temperature range of 2° to 8°C, factory setpoint at 5°C
  • Convenient, sliding drawer baskets for simplified inventory and access
  • Lower HVAC costs as a result of less heat emission into the environment*
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
  • Optional chart recorders available
  • Warranty varies by country, please contact us for details
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.

Figures

Frequently asked questions (FAQs)

Citations & References