TSX Series High-Performance Chromatography Refrigerators
TSX Series High-Performance Chromatography Refrigerators
TSX Series High-Performance Chromatography Refrigerators
TSX Series High-Performance Chromatography Refrigerators
TSX Series High-Performance Chromatography Refrigerators
TSX Series High-Performance Chromatography Refrigerators
Thermo Scientific™

TSX Series High-Performance Chromatography Refrigerators

Innovative refrigeration solutions for healthcare and research needs

Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Door StyleElectrical Requirements
TSX5005CA51.1 cu. ft.Double Glass115V, 60Hz
TSX2305CA23 cu. ft.Single Glass115V, 60Hz
TSX2305CD23 cu. ft.Single Glass208-230V, 50/60Hz
TSX2305CV23 cu. ft.Single Glass208-230V, 50/60Hz
TSX4505CA45.8 cu. ft.Sliding Glass115V, 60Hz
TSX4505CD45.8 cu. ft.Sliding Glass208-230V, 60Hz
TSX4505CV45.8 cu. ft.Sliding Glass208-230V, 50Hz
TSX5005CD51.1 cu. ft.Double Glass208-230V, 60Hz
TSX5005CV51.1 cu. ft.Double Glass208-230V, 50Hz
TSX3005CV29.2 cu. ft.Glass208-230V, 50Hz
TSX3005CY29.2 cu. ft.Glass115 V 60 Hz
TSX3005CZ29.2 cu. ft.Glass208 - 230 V 60 Hz
TSX2305CY23 cu. ft.Glass115 V 60 Hz
TSX2305CZ23 cu. ft.Glass208 - 230 V 50/60 Hz
TSX4505CY45.8 cu. ft.Sliding Glass115 V 60 Hz
TSX4505CZ45.8 cu. ft.Sliding Glass208 - 230 V 60 Hz
TSX5005CY51.1 cu. ft.Double Glass115 V 60 Hz
TSX5005CZ51.1 cu. ft.Double Glass208 - 230 V 60 Hz
Catalog number TSX5005CA
Price (EUR)
-
Capacity (English):
51.1 cu. ft.
Door Style:
Double Glass
Electrical Requirements:
115V, 60Hz

Thermo Scientific™ TSX Series high-performance chromatography refrigerators are designed for a variety of applications requiring precise temperature control, full access to chromatography instrumentation, and easy setup of instrumentation and apparatus within the chamber. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns and offers significant energy savings without compromising protection.

The TSX Chromatography series features industry-leading safety features to ensure maximum protection as well as delivering over 450W of reserve capacity to handle the heat load generated by instruments within the cabinet and to react quickly to frequent door access.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

The drive for sample integrity and security

  • Designed to safely store chromatography and other lab instrumentation requiring a temperature range of 2°C to 8°C
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance
  • Two 2 in. (5 cm) side wall access ports with insulated hinged covers for solvent, waste, and other common tubing and hoses used in chromatography applications.
  • Hydrocarbon Safety switch protected internal Duplex outlet standard on all models

Enabling compliance

  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing glass doors with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life. **
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Alternate Door swing configuration and door types
  • Extra and Alternate wire baskets, shelves, and drawers
  • Optional Chromatography support shelves
  • Free-standing or factory-installed chart recorders
  • Stainless Steel interior finish
  • E-Lock Adapter kits
  • 4-20 milliamp transmitters and other remote alarm modules
  • Specialty, regional, and country specific plugs

Services and support

  • Industry Leading 120-month compressor warranty (varies by country, please contact us for details)
  • Optional Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
  • Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs
Specifications
Amperage15 A
Chart RecorderOptional
DefrostAutomatic
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96 cm
Depth (Metric) Interior72.4 cm
Dimensions (D x W x H) Interior28.5 x 52.5 x 59 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
DisplayMicroprocessor
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Product LineTSX Series
RefrigerantR290
Smart View CompatibilitySmart-Vue Compatible
Temperature (Setpoint)3°-7°C (5°C)
TypeHigh-Performance Chromatography Refrigerator
WarrantyWarranty varies by country, please contact us for details
Width (English) Exterior56.5 in.
Width (English) Interior52.5 in.
Width (Metric) Exterior143.5 cm
Width (Metric) Interior133.4 cm
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Certifications/ComplianceUL, cUL, RoHS
Door StyleDouble Glass
Monitoring OptionsAlarm, Recorder
No. of Doors2
Plug TypeNEMA 5-15
Shelves4 Full, 2 Half
Voltage115 V
Unit SizeEach