Search
Search
| 製品番号(カタログ番号) | 保持 | 容量(ヤードポンド法) | ラック収容量 | 容量(メートル法) |
|---|---|---|---|---|
| TSX50086FA-ULTS | 2 in. Boxes (500) | 24.1立方フィート | 20 | 682 L |
| TSX40086FA | 2インチボックス(400) | 19.4立方フィート | 16 | 549 L |
| TSX50086FA | 2インチボックス(500) | 24.1立方フィート | 20 | 682 L |
| TSX60086FA | 2インチボックス(600) | 28.8立方フィート | 24 | 816 L |
| TSX70086FA | 2インチボックス(700) | 33.5立方フィート | 28 | 949 L |
| TSX40086FA-ULTS | 2 in. Boxes (400) | 19.4立方フィート | 16 | 549 L |
| TSX60086FA-ULTS | 2 in. Boxes (600) | 28.8立方フィート | 24 | 816 L |
| TSX70086FA-ULTS | 2 in. boxes (700) | 33.5 cu. ft. | 28 | 950 L |
TSX Universal—Over 80 years of innovation packed into the next-gen ultra-low freezer. Recognizing the impact of your work, we prioritize sample security and reliability, and our legacy of crafting state-of-the-art lab equipment means our focus remains on user-centric design. The TSX Universal heralds a new era in performance with Universal V-Drive technology, delivering tighter control and faster recovery times. Coupled with an expanded setpoint range, universal voltage, whisper-quiet operation, simplified software updates and much more, TSX Universal seamlessly adapts to your workflow. Redefining sustainability, it is ENERGY STAR† certified with a 32% efficiency improvement, ACT† labeled, and is produced in our Zero-Waste to Landfill† certified facility. TSX Universal ULT—The pinnacle of reliability, sustainability, and innovation.
Reliability at its core:
Built to perform, designed to protect:
Greener by design
Enhanced experience, optimized for ease
*Shipped documents:
Certificate of conformance, CE declaration of conformance, certificate of calibration, QC test log sheets, and QC temperature graphs.
**Restrictions and exclusions apply. Warranty is void if the unit is not maintained as set forth in the operation.
***Zero waste defined as 90% of non-hazardous waste diverted from landfill, waste to energy and incineration.
† Information not applicable to TSX Universal ULT freezers with “-ULTS” in the part number (TSX40086FA-ULTS, TSX50086FA-ULTS, TSX60086FA-ULTS.)