Search
Search
| Catalog Number | Model | Holds | Rack Capacity | Capacity (Metric) |
|---|---|---|---|---|
| TSX70086FA-ULTS | Ultra-Low Freezer 700 | 2 in. boxes (700) | 28 Racks | 950 L |
| TSX40086FA | General Purpose Ultra-Low Freezer 400 | 2 in. Boxes (400) | 16 Racks | 549 L |
| TSX50086FA | General Purpose Ultra-Low Freezer 500 | 2 in. Boxes (500) | 20 Racks | 682 L |
| TSX60086FA | General Purpose Ultra-Low Freezer 600 | 2 in. Boxes (600) | 24 Racks | 816 L |
| TSX70086FA | General Purpose Ultra-Low Freezer 700 | 2 in. Boxes (700) | 28 Racks | 949 L |
| TSX40086FA-ULTS | Ultra-Low Freezer 400 | 2 in. Boxes (400) | 16 Racks | 549 L |
| TSX50086FA-ULTS | Ultra-Low Freezer 500 | 2 in. Boxes (500) | 20 Racks | 682 L |
| TSX60086FA-ULTS | Ultra-Low Freezer 600 | 2 in. Boxes (600) | 24 Racks | 816 L |
TSX Universal—Over 80 years of innovation packed into the next-gen ultra-low freezer. Recognizing the impact of your work, we prioritize sample security and reliability, and our legacy of crafting state-of-the-art lab equipment means our focus remains on user-centric design. The TSX Universal heralds a new era in performance with Universal V-Drive technology, delivering tighter control and faster recovery times. Coupled with an expanded setpoint range, universal voltage, whisper-quiet operation, simplified software updates and much more, TSX Universal seamlessly adapts to your workflow. Redefining sustainability, it is ENERGY STAR† certified with a 32% efficiency improvement, ACT† labeled, and is produced in our Zero-Waste to Landfill† certified facility. TSX Universal ULT—The pinnacle of reliability, sustainability, and innovation.
Reliability at its core:
Built to perform, designed to protect:
Greener by design
Enhanced experience, optimized for ease
*Shipped documents:
Certificate of conformance, CE declaration of conformance, certificate of calibration, QC test log sheets, and QC temperature graphs.
**Restrictions and exclusions apply. Warranty is void if the unit is not maintained as set forth in the operation.
***Zero waste defined as 90% of non-hazardous waste diverted from landfill, waste to energy and incineration.
† Information not applicable to TSX Universal ULT freezers with '-ULTS' in the part number (TSX40086FA-ULTS, TSX50086FA-ULTS, TSX60086FA-ULTS.)