TSQ Quantis™ Plus Triple Quadrupole MS eLearning Courses
Thermo Scientific™

TSQ Quantis™ Plus Triple Quadrupole MS eLearning Courses

TSQ Quantis™ Plus Triple Quadrupole MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Número de catálogoCourse Topic
TTSQNPMTSMantenimiento por parte del usuario y solución de problemas
Número de catálogo TTSQNPMTS
Precio (MXN)
-
Course Topic:
Mantenimiento por parte del usuario y solución de problemas

TSQ Quantis™ Triple Plus Quadrupole MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The TSQ Quantis™ Plus Triple Quadrupole MS foundational theory eLearning course introduces how triple-stage quadrupole mass spectrometers work, the basic hardware configuration of TSQ Series II systems, and the different scan types they can perform.

Some of the concepts and topics presented include:

  • Principles of MS/MS operation
  • Specificity and sensitivity of mass spectrometers
  • Basic hardware configuration
  • Overview of the vacuum system
  • Components inside the mass spectrometer
  • Scan types

This Foundational Theory eLearning course is similar and applicable to all TSQ Series II instruments; e.g. TSQ Quantiva™, TSQ Endura™, TSQ Quantis™, TSQ Fortis™, TSQ Altis™, TSQ Altis™ Plus, and TSQ Fortis™ Plus.

The TSQ Quantis™ Plus Triple Quadrupole MS user maintenance and troubleshooting eLearning course provides detailed instructions through video demonstrations of several routine maintenance procedures performed on the Thermo Scientific™ TSQ™ Series II triple quadrupole mass spectrometers with an OptaMax ion source.

  • Removal, disassembly, cleaning, reassembly, and reinstallation of the external components of the API source and OptaMax source.
  • Removal, disassembly, cleaning, reassembly, and reinstallation of the external components of the API source interface lenses.
  • Replacement of the H-ESI spray insert needle.
  • Replacement of the ESI spray insert needle.
  • Replacement of the APCI spray insert sample tube.
  • Replacement of the APCI corona needle.

The content included in this training module applies to the second and third-generation TSQ™ Series II instruments: Altis™, Quantis™, Fortis™, Altis™ Plus, Quantis™ Plus, and Fortis™ Plus.

Especificaciones
Access Availablity30 days upon end user first acess
Course TopicMantenimiento por parte del usuario y solución de problemas
FormatoSelf-paced eLearning
IdiomasEnglish
Course LevelRoutine
Duración60 to 120 min.
Para utilizar con (equipo)TSQ Quantis Plus Triple Quadrupole MS
Tipo de productoMass Spectrometry
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach