Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips
Imagem ilustrativa. O produto pode variar
Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips
Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips
Thermo Scientific™

Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips

Thermo Scientific™ QSP™ Thin Wall Micro Point Pipette Tips help deliver samples with a nice uniform bead, cleanly breaking liquid surface tension easily releasing liquid from the distal tip end.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoSterilityFormat
TTW110RLNS-QSterileReload System
TTW110RLN-QNon-sterileReload System
TTW110RL-QNon-sterileReload System
TTW110RN-QNon-sterileRack
TTW110RLS-QSterileReload System
TTW110R-QNon-sterileRack
TW110-QNon-sterileBulk Bags
TW110-96RNS-QNon-sterileReload System
Número do catálogo TTW110RLNS-Q
Preço (BRL)
-
Sterility:
Sterile
Format:
Reload System

Thermo Scientific™ QSP™ Thin Wall Micro Point 200μl natural pipette tips. Packaged in reload system and non-sterile clean claims are; Rnase/Dnase, DNA, pyrogen and endotoxin free. Universal fit on most pipettes.

  • Thin wall micropoint design decreases surface tension helping liquid bead up cleanly
  • Great for plating
  • Yellow and natural in a variety of packaging options
Especificações
Clean ClaimsSterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden
AutoclavableAutoclavable
ColorNatural
For Use With (Equipment)Universal Fit
FormatReload System
Material100% Virgin Polypropylene
Product LineQSP
SterilitySterile
Volume (Metric)200 μL
Unit SizeCase of 4800