Oxoid™ Prepared Brilliant Green Bile Broth D/T Bottled Media
Thermo Scientific™

Oxoid™ Prepared Brilliant Green Bile Broth D/T Bottled Media

A selective broth for the detection of coliforms in water and other samples.
Have Questions?
變更視圖buttonViewtableView
產品號碼Fill WeightFormat
TV5009E9.5 to 10.5 gPoured Tube
BO0345E10.0-10.3g Universal bottle
產品號碼 TV5009E
價格 (TWD)
-
Fill Weight:
9.5 to 10.5 g
Format:
Poured Tube
For the detection of coliforms in water and other samples.
規格
ColorMint Green, Transparent
Fill Weight9.5 to 10.5 g
FormatPoured Tube
Packaging Type50 Tubes/Box
Quantity50/Pk.
Shelf Life26 weeks
Manufacturing OriginGermany
Product TypeMicrobiology Media
Unit SizeEach
內容物與存放
2°C to 25°C