Differentiate enteric bacteria based on lysine decarboxylation, hydrogen sulfide (H2S) production, and glucose fermentation.
Have Questions?
Número de catálogo
Peso de llenado
TV5045D
De 8,5 a 9,5 g
Número de catálogo TV5045D
Precio (MXN)
-
Solicite un presupuesto
Peso de llenado:
De 8,5 a 9,5 g
Lysine Iron Slant Agar is used to differentiate enteric bacteria based on lysine decarboxylation, hydrogen sulfide (H2S) production, and glucose fermentation.