Vanquish™ Fluorescence Detector Maintenance and Troubleshooting
Thermo Scientific™

Vanquish™ Fluorescence Detector Maintenance and Troubleshooting

The eLearning courses for the Vanquish™ Fluorescence Detectors provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses that can cover different topics such as foundational theory, basic operations principles, and/or user maintenance and troubleshooting.
Have Questions?
Catalog NumberCourse TopicCourse Level
TVFLDMTSUser Maintenance and TroubleshootingRoutine
Catalog number TVFLDMTS
Price (EUR)
-
Course Topic:
User Maintenance and Troubleshooting
Course Level:
Routine

The eLearning courses for the Vanquish™ Fluorescence Detectors provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses that can cover different topics such as foundational theory, basic operations principles, and/or user maintenance and troubleshooting.

The Vanquish™ Fluorescence Detectors (FLD) User Maintenance and Troubleshooting eLearning course provides instruction and video demonstrations on maintenance procedures and troubleshooting tips for the Thermo Scientific™ Vanquish™ fluorescence detectors.Topics covered in this presentation include:

  • General maintenance rules and plans
  • Monitoring lamp age
  • Wavelength calibration and validation
  • Flow cell maintenance

Troubleshooting tips:

  • LED indicators
  • Audit trail messages
  • Liquid leaks
  • Additional operational issues
Specifications
Access Availablity30 days upon end user first access to the course
Course TopicUser Maintenance and Troubleshooting
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelRoutine
Duration<30 min.
For Use With (Equipment)Vanquish Fluorescence Detector
Product LineThermo Scientific
Product TypeChromatography
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach