ULYSIS™ Alexa Fluor™ Nucleic Acid Labeling Kit
Invitrogen™

ULYSIS™ Alexa Fluor™ Nucleic Acid Labeling Kit

Achieve unparalleled precision and sensitivity in your fluorescence-based applications from FISH to real-time PCR. With a streamlined, non-enzymatic labeling process and a broad selection of vibrant dyes, ULYSIS kits ensure your nucleic acids are ready for any challenge.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호라벨 또는 염료여기/방출
U21652Alexa Fluor 546555/570 nm
U21650Alexa Fluor 488492/520 nm
U21654Alexa Fluor 594588/615 nm
U21660Alexa Fluor 647650/670 nm
카탈로그 번호 U21652
제품 가격(KRW)
783,000
온라인 행사
Ends: 31-Dec-2026
921,000
할인액 138,000 (15%)
1 kit
라벨 또는 염료:
Alexa Fluor 546
여기/방출:
555/570 nm
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
783,000
온라인 행사
Ends: 31-Dec-2026
921,000
할인액 138,000 (15%)
1 kit

ULYSIS nucleic acid labeling kits provide a unique method for attaching a fluorescent dye to nucleic acids. The labeling reagent in the kit reacts with the N7 of guanine to form a stable coordination complex, and the reaction is simple and fast - just heat denature DNA (5 minutes), add the label (react for 15 minutes), then purify.

Features include:

  • Labeling reaction complete in as little as 15 minutes
  • Available in several Alexa Fluor dye colors
  • Non-enzymatic labeling—avoids the potential biases and limitations associated with enzymatic labeling methods
  • Flexibility—compatible with a wide range of experimental conditions and protocols
  • High sensitivity—provides strong fluorescence signals, enabling detection of low-abundance targets

Reliable labeling with the Universal Linkage System

This series of ULYSIS kits was developed to enable rapid and simple coupling of Alexa Fluor dyes to purine bases in nucleic acid polymers. The method, the Universal Linkage System (ULS), makes use of a platinum dye complex that forms a stable adduct with the N7 position of guanine and, to a lesser extent, adenine bases in DNA, RNA, PNA, and oligonucleotides. The result is a reliable nonenzymatic method for nucleic acid labeling.

Labeling is fast and easy

The labeling reaction typically takes only 15 minutes, and separation of the labeled nucleic acids from the unreacted ULS complex can be accomplished with a simple spin-column procedure. DNA longer than ∼1,000 base pairs require a 10-minute DNase digestion before labeling, which both optimizes labeling and fragments the probe for efficient hybridization.

For Research Use Only. Not for use in diagnostic procedures.
사양
색상Orange
여기/방출555/570 nm
용도(애플리케이션)Dot blot, Northern blot, Southern blot, RNA in situ hybridization, DNA in situ hybridization, Multicolor fluorescence in situ hybridization (mFISH), Comparative genome hybridization (CGH), Microarray analysis
라벨 또는 염료 포함Yes
라벨링 방법Direct Labeling
제품라인Alexa Fluor, Ulysis
제품 유형Nucleic Acid Labeling Kit
수량1 Kit
배송 조건Room Temperature
검출 방법Fluorescence
최종 제품 유형Probes (Labeled RNA), Probes (Labeled DNA), Oligos (Labeled)
형식Kit
Labeling TargetDNA (General), Oligos, RNA (General)
라벨 또는 염료Alexa Fluor 546
샘플 종류DNA/RNA
Unit Size1 kit
구성 및 보관
Store in freezer (-5 to -30°C) and protect from light.

Needed but not included: gel filtration-based spin columns for purification of the labeled DNA from excess labeling reagent

자주 묻는 질문(FAQ)

Is ULYSIS labeling compatible with microarray analysis?

Yes, there are numerous examples of ULYSIS labeled probes that have been used in microarray analysis. Here are a few publications for your reference:

- Babak T, Zhang W, Marros Q et al. (2004) Probing microRNAs with microarrays: tissue specificity and functional inference. RNA 10(11):1813-1819.
- Hiley SL, Jackman J, Babak T et al. (2005) Detection and discovery of RNA modifications using microarrays. Nucleic Acids Res 33(1):e2.
- Torchet C, Badis G, Devaux F et al. (2005) The complete set of H/ACA snoRNAs that guide rRNA pseudouridylations in Saccharomyces cerevisiae. RNA 11(6):928-938.

Can probes labeled with the ULYSIS Nucleic Acid Labeling Kits be stored for later use?

Long-term storage for the ULYSIS labeled probes can be done in just about any kind of buffer, TE, formamide, hybridization buffer, or ethanol. We suggest using your normal storage conditions as long as you protect the probes from light. ULYSIS conjugates are very stable. Avoid phenol.

Do you have any tips on using the ULYSIS Nucleic Acid Labeling Kits for RNA labeling?

A preliminary protocol modifies our DNA-labeling protocol: Do not nuclease-treat the RNA, but label it directly by incubating for 10 minutes at 90°C or 15 minutes at 85°C. Add 2 µg of glycogen for every 1 µg of RNA and purify by ethanol precipitation. Refer to these publications:

- Babak T, Zhang W, Marros Q et al. (2004) Probing microRNAs with microarrays: tissue specificity and functional inference. RNA 10(11):1813-1819.
- Hiley SL, Jackman J, Babak T et al. (2005) Detection and discovery of RNA modifications using microarrays. Nucleic Acids Res 33(1):e2.
- Torchet C, Badis G, Devaux F et al. (2005) The complete set of H/ACA snoRNAs that guide rRNA pseudouridylations in Saccharomyces cerevisiae. RNA 11(6):928-938.

Can the ULYSIS kits be used on probes longer than 1,000 base pairs or even plasmids?

It might be possible to label larger probes with the ULYSIS Nucleic Acid Labeling Kits, but the dye will likely need to be diluted to avoid (or at least reduce) problems with aggregation. Refer to this publication: Coelho-Castelo AA, Santos Junior RR, Bonato VL et al. (2003) B-lymphocytes in bone marrow or lymph nodes can take up plasmid DNA after intramuscular delivery. Hum Gene Ther 14(13):1279-1285.

How stable is the ULYSIS labeled DNA to high temperature?

An oligonucleotide labeled with a ULYSIS Nucleic Acid Labeling Kit should survive 100°C for 5 minutes, and storage at 68°C overnight should also not cause any dissociation of the complex.

인용 및 참조 문헌 (5)

인용 및 참조 문헌
Abstract
A systematic search for new mammalian noncoding RNAs indicates little conserved intergenic transcription.
Authors:Babak T, Blencowe BJ, Hughes TR
Journal:BMC Genomics
PubMed ID:16083503
'BACKGROUND: Systematic identification and functional characterization of novel types of noncoding (nc)RNA in genomes is more difficult than it is for protein coding mRNAs, since ncRNAs typically do not possess sequence features such as splicing or translation signals, or long open reading frames. Recent "tiling" microarray studies have reported that ... More
Expression profile analysis of microRNA (miRNA) in mouse central nervous system using a new miRNA detection system that examines hybridization signals at every step of washing.
Authors:Hohjoh H, Fukushima T
Journal:Gene
PubMed ID:17229533
'MicroRNAs (miRNAs) are small noncoding RNAs, with a length of 19 to 23 nucleotides, which appear to be involved in the regulation of gene expression by inhibiting the translation of messenger RNA. Expression profile analysis of miRNAs is necessary to understand their complex role in the regulation of gene expression ... More
Probing microRNAs with microarrays: tissue specificity and functional inference.
Authors:Babak T, Zhang W, Morris Q, Blencowe BJ, Hughes TR
Journal:RNA
PubMed ID:15496526
MicroRNAs (miRNAs) are short, stable, noncoding RNAs involved in post-transcriptional gene silencing via hybridization to mRNA. Few have been thoroughly characterized in any species. Here, we describe a method to detect miRNAs using micro-arrays, in which the miRNAs are directly hybridized to the array. We used this method to analyze ... More
The clustering of four subfamilies of satellite DNA at individual chromosome ends in Silene latifolia.
Authors:Kazama Y, Sugiyama R, Suto Y, Uchida W, Kawano S
Journal:Genome
PubMed ID:16767177
The satellite DNA (satDNA) on the ends of chromosomes has been isolated and characterized in the dioecious plant Silene latifolia. BAC clones containing large numbers of repeat units of satDNA in a tandem array were isolated to examine the clustering of the repeat units. satDNA repeat units were purified from ... More
Evaluation of cadmium-induced transcriptome alterations by three color cDNA labeling microarray analysis on a T-cell line.
Authors:Tsangaris GT, Botsonis A, Politis I, Tzortzatou-Stathopoulou F
Journal:Toxicology
PubMed ID:12160620
Beside heavy metals, cadmium (Cd(2+)) is a ubiquitous toxic metal with a well established apoptotic and genotoxic effect, chronic exposure of which has been involved in a variety of pathological conditions. In the present study, we investigated by 1455 genes cDNA microarrays the toxic and apoptotic effect of Cd(2+), on ... More