MAS™ UA DipTube Control
MAS™ UA DipTube Control
Thermo Scientific™

MAS™ UA DipTube Control

Simplify QC during urinalysis testing with room-temperature, stable Thermo Scientific™ MAS™ UA DipTube Control, conveniently packed for time and cost savings.
Have Questions?
Catalog NumberQuantity
UAT-MP12 mL
Catalog number UAT-MP
Price (USD)
-
Request A Quote
Quantity:
12 mL
Simplify the process of quality control during urinalysis testing with room-temperature, stable Thermo Scientific™ MAS™ UA DipTube Control. This liquid assayed urinalysis control for procedures used in routine urinalysis testing is conveniently-packed to afford immediate time and cost savings.
Features:
  • Key urinalysis test parameters provided in a single control
  • Tube capacity common within urinalysis test environment
  • Assayed for most major reagent strips and strip systems
  • Human urine matrix

Ordering Information:

Packaged in 12mL dip tube format; 5 vials of each level (1 and 2)

Requires:

  • Slides
  • Pipetters
  • Centrifuge capable of generating an RCF appropriate for the microscopic method used by the laboratory
  • Microscope with a 10x objective and a 40x objective lens
  • Chemical reagent strips
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionAssayed, Bi-level Multi-Pack; 10 x 12mL
LevelBi-level
Packaging10 Pk.
Quantity12 mL
Shelf Life24 months
TypeControl
Detectable AnalytesBilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C