Search
| Número do catálogo | For Use With (Application) | Emission Spectra |
|---|---|---|
| UCW11727 | Environmental, Personnel, Extremity Dosimetry | 4000 Å |
| UCW71727 | Gamma, Beta and Environmental | 4000 Å |
| UNW78020 | Gamma and Beta Dosimetry | 3500 to 6000 Å (4000 maximum) |
| UNW28020 | Environmental Dosimetry | 4835 Å |
| UNW68020 | Neutron Dosimetry | 3500 to 6000 Å (4000 maximum) |
| UCW61727 | Neutron Dosimetry | 4000 Å |
| UNW18020 | Health and Medical Physics Dosimetry | 3500 to 6000 Å (4000 maximum) |
Get accurate radiation dose evaluation for applications from health physics to environmental to personnel and extremity, and beyond with highly sensitive thermoluminescent dosimetry powders. Thermo Scientific™ Thermoluminescent Dosimetry Powders are accurate for x-ray radiation, gamma radiation, beta radiation, neutron radiation, and electron radiation according to choice of material.
TLD-600 materials consist of Lithium Fluoride (Li isotope) LiF:Mg, Ti; suitable for gamma and beta dosimetry applications.