Thermoluminescent Dosimetry Powders
Actual product may vary
Thermo Scientific™

Thermoluminescent Dosimetry Powders

Get accurate radiation dose measurement for any application, from health physics to environmental dosimetry applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)Emission Spectra
UCW71727Gamma, Beta and Environmental4000 Å
UNW78020Gamma and Beta Dosimetry3500 to 6000 Å (4000 maximum)
UNW28020Environmental Dosimetry4835 Å
UNW68020Neutron Dosimetry3500 to 6000 Å (4000 maximum)
UCW11727Environmental, Personnel, Extremity Dosimetry4000 Å
UCW61727Neutron Dosimetry4000 Å
UNW18020Health and Medical Physics Dosimetry3500 to 6000 Å (4000 maximum)
Catalog number UCW71727
Price (MXN)
-
For Use With (Application):
Gamma, Beta and Environmental
Emission Spectra:
4000 Å

Get accurate radiation dose evaluation for applications from health physics to environmental to personnel and extremity, and beyond with highly sensitive thermoluminescent dosimetry powders. Thermo Scientific™ Thermoluminescent Dosimetry Powders are accurate for x-ray radiation, gamma radiation, beta radiation, neutron radiation, and electron radiation according to choice of material.

TLD-600 materials consist of Lithium Fluoride (Li isotope) LiF:Mg, Ti; suitable for gamma and beta dosimetry applications.


Specifications
For Use With (Application)Gamma, Beta and Environmental
Measurement Ranges1 pGy to 10 Gy
Radiation TypeBeta, Gamma
Emission Spectra4000 Å
Energy Response0.98
MaterialLithium Fluoride (Li isotope) LiF:Mg, Cu, P
Product TypeThermoluminescent Dosimetry Material
Sensitivity15
Unit SizeEach