Revco™ Enzyme Freezer
Revco™ Enzyme Freezer
Revco™ Enzyme Freezer
Revco™ Enzyme Freezer
Thermo Scientific™

Revco™ Enzyme Freezer

Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Plug TypeVoltage
UEN3020D29.2 cu. ft.NEMA 6-15 P208/230 V
UEN2320A23.3 cu. ft.NEMA 5-20 P115 V
UEN2320D23.3 cu. ft.NEMA 6-15 P208/230 V
UEN2320V23.3 cu. ft.EU230 V
UEN2320W23.3 cu. ft.UK230 V
UEN3020A29.2 cu. ft.NEMA 5-15 P115 V
UEN3020V29.2 cu. ft.EU230 V
UEN3020W29.2 cu. ft.UK230 V
Catalog number UEN3020D
Price (MXN)
-
Capacity (English):
29.2 cu. ft.
Plug Type:
NEMA 6-15 P
Voltage:
208/230 V

Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired. Our manual defrost -20°C upright freezers feature microprocessor control system, gravity air circulation, industrial-quality cabinet construction and extra-strength refrigeration compressors designed for industrial, clinical and scientific use.

Key Features
  • Operating temperature: -20°C
  • Preset temperature setpoint
  • Upright 23.3 cu. ft. (659L) capacity
  • Gravity convection air system for maximum stability without blowers or fans
  • Manual defrost

Microprocessor Control System:

  • Icon-based controls and displays
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
  • Large, easy-to-read digital display of temperature within 0.1°C
  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Battery backup system

Construction:

  • Scratch-resistant, painted cold-rolled steel interior and exterior finishes
  • Solid door with full-length handle and keyed lock—self-closes against closed-cell peripheral gasket
  • High-density, CFC-free urethane foam insulation protects cabinet from ambient temperature fluctuations and minimizes operating costs
  • Hermetically sealed heavy-duty, industrial-grade compressor
  • Nine epoxy-coated, open-wire shelves, adjustable in 1in. (2.5cm) increments
  • Adjustable leveling feet

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors available separately. Contact your Customer Service or Sales Representative for details.

Includes:

45 high-impact plastic enzyme storage bins for easily accessible inventory. Bin overall dimensions (L × W × H): 25.25 × 4.25 × 3in. (64 × 11 × 8cm); labels not provided.

  • Warranty varies by country, please contact us for details
  • Specifications
    Amperage5.6 A
    Breaker15A
    Control TypeMicroprocessor
    DefrostManual
    Depth (English) Interior29 in.
    Depth (Metric) Interior74 cm
    Dimensions (D x W x H) Interior29 x 30 x 58 in. (74 x 76 x 147 cm)
    Dimensions (L x W x H) Exterior37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201 cm)
    Electrical Requirements208 - 230 V 60 Hz
    Exterior FinishEpoxy Coated Steel
    Frequency60 Hz
    Height (English) Exterior79.2 in.
    Height (English) Interior58 in.
    Height (Metric) Exterior201 cm
    Height (Metric) Interior147 cm
    InsulationUrethane Foam
    Length (English) Exterior37.2 in.
    Length (Metric) Exterior94.5 cm
    Rack Capacity9
    RefrigerantCFC-free
    Shipping Weight (English)445 lb.
    Shipping Weight (Metric)197 kg
    TypeEnzyme Freezer
    WarrantyWarranty varies by country, please contact us for details
    Width (English) Exterior34 in.
    Width (English) Interior30 in.
    Width (Metric) Exterior86.4 cm
    Width (Metric) Interior76 cm
    Cabinet MaterialScratch-resistant Painted
    Capacity (English)29.2 cu. ft.
    Capacity (Metric)826 L
    Certifications/ComplianceUL
    CompressorHeavy-duty, Industrial-grade, Hermetic
    Door StyleSolid
    Monitoring OptionsAlarm, Recorder
    No. of Doors1
    Plug TypeNEMA 6-15 P
    Shelves9
    StyleUpright
    Temperature Range-20°C
    Voltage208/230 V
    Unit SizeEach