Revco™ High-Performance Manual Defrost Lab Freezers
Revco™ High-Performance Manual Defrost Lab Freezers
Revco™ High-Performance Manual Defrost Lab Freezers
Revco™ High-Performance Manual Defrost Lab Freezers
Thermo Scientific™

Revco™ High-Performance Manual Defrost Lab Freezers

Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)VoltagePlug Type
UGL3020D29.2 cu. ft.208/230 VNEMA 6-15 P
UGL2320A23.3 cu. ft.115 VNEMA 5-20 P
UGL2320D23.3 cu. ft.208/230 VNEMA 6-15 P
UGL2320V23.3 cu. ft.230 VEU
UGL2320W23.3 cu. ft.230 VUK
UGL3020A29.2 cu. ft.115 VNEMA 5-20 P
UGL3020V29.2 cu. ft.230 VEU
UGL3020W29.2 cu. ft.230 VUK
Catalog number UGL3020D
Price (MXN)
-
Capacity (English):
29.2 cu. ft.
Voltage:
208/230 V
Plug Type:
NEMA 6-15 P
Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers. Our Revco™ -30°C and -20°C high-performance freezers are designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals or other commonly used laboratory materials. Models offer cabinet sizes to fit a variety of space needs and storage volume requirements.
Key Features
  • Temperature factory-preset at -20°
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature

Microprocessor Control

  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system

Construction

  • Scratch-resistant cold-rolled steel interior and exterior finishes
  • High-density, 2 in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
  • CFC-free, HFC-blend refrigerant
  • Single-door models are hinged on right
  • Full-length door handle and key lock
  • Doors self-close and lock open at 90°
  • Adjustable epoxy-coated wire shelves
  • 1 in. (2.5cm) dia. access port with plug on undercounter model

Model Specific Features:

-20°C Freezers

  • Suitable for applications in which slight intermittent coil warming during an automatic defrost cycle is not desired (such as enzyme storage)
  • Gravity convection air system for maximum stability without a blower or fan
  • Manual defrost

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors are available separately.

Note:

Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative.

Warranty varies by country, please contact us for details
Specifications
Amperage12 A
Breaker15A
Cabinet MaterialCold-rolled Steel
Control TypeMicroprocessor
DefrostManual
Depth (English) Interior29 in.
Depth (Metric) Interior74 cm
Dimensions (D x W x H) Interior29 x 30 x 58 in. (74 x 76 x 147 cm)
Dimensions (L x W x H) Exterior37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201 cm)
Door StyleSolid
Electrical Requirements208 - 230 V 60 Hz
Exterior FinishEpoxy Coated Steel
Frequency60 Hz
Height (English) Exterior79.2 in.
Height (English) Interior58 in.
Height (Metric) Exterior201 cm
Height (Metric) Interior147 cm
InsulationUrethane Foam
Length (English) Exterior37.2 in.
Length (Metric) Exterior94.5 cm
Plug TypeNEMA 6-15 P
RefrigerantCFC-free
Shipping Weight (English)446 lb.
Shipping Weight (Metric)197 kg
TypeHigh-Performance Freezer
WarrantyWarranty varies by country, please contact us for details
Width (English) Exterior34 in.
Width (English) Interior30 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior76 cm
Capacity (English)29.2 cu. ft.
Capacity (Metric)826 L
Certifications/ComplianceUL
Monitoring OptionsAlarm, Recorder
No. of Doors1
Shelves4
StyleUpright
Temperature Range-20°C
Voltage208/230 V
Unit SizeEach