Thermo Scientific™

Revco™ High-Performance Auto Defrost Lab Freezers

Catalog number: ULT430D
Thermo Scientific™

Revco™ High-Performance Auto Defrost Lab Freezers

Catalog number: ULT430D
Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers. Our Revco™ -30°C and -20°C high-performance freezers are designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals or other commonly used laboratory materials. Models offer cabinet sizes to fit a variety of space needs and storage volume requirements.
 
Catalog Number
ULT430D
Unit Size
Each
Voltage
208/230 V
Plug Type
NEMA 6-15 P
Temperature Range
-30°C
Capacity (English)
4.9 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)4.9 cu. ft.
Capacity (Metric)139 L
Temperature Range-30°C
Door StyleSolid
No. of Doors1
Cabinet MaterialCold-rolled Steel
InsulationUrethane Foam
Shelves3
Certifications/ComplianceCE, cUL, UL
StyleUndercounter
Control TypeMicroprocessor
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior20.5 x 20 x 20 in. (52.1 x 50.8 x 50.8 cm)
Dimensions (L x W x H) Exterior28.9 x 24 x 34 in. (73.4 x 61 x 86.4 cm)
Depth (English) Interior20 in.
Depth (Metric) Interior50.8 cm
Height (English) Exterior34 in.
Height (English) Interior20.5 in.
Height (Metric) Exterior86.4 cm
Height (Metric) Interior52.1 cm
Length (English) Exterior28.9 in.
Length (Metric) Exterior73.4 cm
Width (English) Exterior24 in.
Width (English) Interior20 in.
Width (Metric) Exterior61 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsAlarm, Recorder
Amperage6 A
Breaker15A
Plug TypeNEMA 6-15 P
Voltage208/230 V
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
RefrigerantCFC-free
Shipping Weight (English)212 lb.
Shipping Weight (Metric)96 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Voltage
115 V
Plug Type
NEMA 5-15 P
Temperature Range
-30°C
Capacity (English)
4.9 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)4.9 cu. ft.
Capacity (Metric)139 L
Temperature Range-30°C
Door StyleSolid
No. of Doors1
Cabinet MaterialCold-rolled Steel
InsulationUrethane Foam
Shelves3
Certifications/ComplianceCE, cUL, UL
StyleUndercounter
Control TypeMicroprocessor
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior20.5 x 20 x 20 in. (52.1 x 50.8 x 50.8 cm)
Dimensions (L x W x H) Exterior28.9 x 24 x 34 in. (73.4 x 61 x 86.4 cm)
Depth (English) Interior20 in.
Depth (Metric) Interior50.8 cm
Height (English) Exterior34 in.
Height (English) Interior20.5 in.
Height (Metric) Exterior86.4 cm
Height (Metric) Interior52.1 cm
Length (English) Exterior28.9 in.
Length (Metric) Exterior73.4 cm
Width (English) Exterior24 in.
Width (English) Interior20 in.
Width (Metric) Exterior61 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsAlarm, Recorder
Amperage8 A
Breaker15A
Plug TypeNEMA 5-15 P
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
RefrigerantCFC-free
Shipping Weight (English)212 lb.
Shipping Weight (Metric)96 kg
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeVoltagePlug TypeTemperature RangeCapacity (English)Price (USD)
ULT430DFull specifications
Each208/230 VNEMA 6-15 P-30°C4.9 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)4.9 cu. ft.
Capacity (Metric)139 L
Temperature Range-30°C
Door StyleSolid
No. of Doors1
Cabinet MaterialCold-rolled Steel
InsulationUrethane Foam
Shelves3
Certifications/ComplianceCE, cUL, UL
StyleUndercounter
Control TypeMicroprocessor
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior20.5 x 20 x 20 in. (52.1 x 50.8 x 50.8 cm)
Dimensions (L x W x H) Exterior28.9 x 24 x 34 in. (73.4 x 61 x 86.4 cm)
Depth (English) Interior20 in.
Depth (Metric) Interior50.8 cm
Height (English) Exterior34 in.
Height (English) Interior20.5 in.
Height (Metric) Exterior86.4 cm
Height (Metric) Interior52.1 cm
Length (English) Exterior28.9 in.
Length (Metric) Exterior73.4 cm
Width (English) Exterior24 in.
Width (English) Interior20 in.
Width (Metric) Exterior61 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsAlarm, Recorder
Amperage6 A
Breaker15A
Plug TypeNEMA 6-15 P
Voltage208/230 V
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
RefrigerantCFC-free
Shipping Weight (English)212 lb.
Shipping Weight (Metric)96 kg
Unit SizeEach
ULT430AFull specifications
Each115 VNEMA 5-15 P-30°C4.9 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)4.9 cu. ft.
Capacity (Metric)139 L
Temperature Range-30°C
Door StyleSolid
No. of Doors1
Cabinet MaterialCold-rolled Steel
InsulationUrethane Foam
Shelves3
Certifications/ComplianceCE, cUL, UL
StyleUndercounter
Control TypeMicroprocessor
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior20.5 x 20 x 20 in. (52.1 x 50.8 x 50.8 cm)
Dimensions (L x W x H) Exterior28.9 x 24 x 34 in. (73.4 x 61 x 86.4 cm)
Depth (English) Interior20 in.
Depth (Metric) Interior50.8 cm
Height (English) Exterior34 in.
Height (English) Interior20.5 in.
Height (Metric) Exterior86.4 cm
Height (Metric) Interior52.1 cm
Length (English) Exterior28.9 in.
Length (Metric) Exterior73.4 cm
Width (English) Exterior24 in.
Width (English) Interior20 in.
Width (Metric) Exterior61 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsAlarm, Recorder
Amperage8 A
Breaker15A
Plug TypeNEMA 5-15 P
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
RefrigerantCFC-free
Shipping Weight (English)212 lb.
Shipping Weight (Metric)96 kg
Unit SizeEach
Showing 2 of 2
Key Features
  • Temperature factory-preset at -30°C
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature

Microprocessor Control

  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system

Construction

  • Scratch-resistant cold-rolled steel interior and exterior finishes
  • High-density, 2 in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
  • CFC-free, HFC-blend refrigerant
  • Single-door models are hinged on right
  • Full-length door handle and key lock
  • Doors self-close and lock open at 90°
  • Adjustable epoxy-coated wire shelves
  • 1 in. (2.5cm) dia. access port with plug on undercounter model

Model Specific Features:

-30°C Freezers

  • Automatic defrost and condensate removal
  • Sensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
  • Forced-air circulation improves temperature uniformity, provides faster recovery after door is opened

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors are available separately.

Note:

Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative.

Warranty varies by country, please contact us for details

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text