Revco™ High-Performance Auto Defrost Lab Freezers
Actual product may vary
Thermo Scientific™

Revco™ High-Performance Auto Defrost Lab Freezers

Simplify operation and increase accuracy with the microprocessor control of these high-performance lab freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)VoltagePlug Type
ULT5030V51.1 cu. ft.230 VEU
ULT430A4.9 cu. ft.115 VNEMA 5-15 P
ULT430W4.9 cu. ft.230 VUK
ULT1230A11.5 cu. ft.115 VNEMA 5-15 P
ULT1230V11.5 cu. ft.230 VEU
ULT1230W11.5 cu. ft.230 VUK
ULT2330A23.3 cu. ft.115 VNEMA 5-20 P
ULT2330D23.3 cu. ft.208/230 VNEMA 6-15
ULT3030A29.2 cu. ft.115 VNEMA 5-20 P
ULT3030D29.2 cu. ft.208/230 VNEMA 6-15
ULT3030V29.2 cu. ft.220 VNEMA 5-20 P
ULT3030W29.2 cu. ft.230 VUK
ULT5030A51.1 cu. ft.115 VNEMA 5-20 P
ULT5030D51.1 cu. ft.208/230 VNEMA 6-15 P
ULT5030W51.1 cu. ft.230 VUK
Catalog number ULT5030V
Price (MXN)
-
Capacity (English):
51.1 cu. ft.
Voltage:
230 V
Plug Type:
EU
Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers. Our Revco™ -30°C and -20°C high-performance freezers are designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals or other commonly used laboratory materials. Models offer cabinet sizes to fit a variety of space needs and storage volume requirements.
Key Features
  • Temperature factory-preset at -30°C
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature

Microprocessor Control

  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system

Construction

  • Scratch-resistant cold-rolled steel interior and exterior finishes
  • High-density, 2 in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
  • CFC-free, HFC-blend refrigerant
  • Single-door models are hinged on right
  • Full-length door handle and key lock
  • Doors self-close and lock open at 90°
  • Adjustable epoxy-coated wire shelves
  • 1 in. (2.5cm) dia. access port with plug on undercounter model

Model Specific Features:

-30°C Freezers

  • Automatic defrost and condensate removal
  • Sensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
  • Forced-air circulation improves temperature uniformity, provides faster recovery after door is opened

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors are available separately.

Note:

Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative.

Warranty varies by country, please contact us for details
Specifications
Amperage12 A
Breaker15A
Cabinet MaterialCold-rolled Steel
Control TypeMicroprocessor
DefrostAutomatic
Depth (English) Interior29 in.
Depth (Metric) Interior73.7 cm
Dimensions (D x W x H) Interior29 x 52.5 x 58 in. (73.7 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior37.2 x 56.5 x 79.2 in. (94.5 x 143 x 201.2 cm)
Door StyleSolid
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Height (English) Exterior79.2 in.
Height (English) Interior58 in.
Height (Metric) Exterior201.2 cm
Height (Metric) Interior147.3 cm
InsulationUrethane Foam
Length (English) Exterior37.2 in.
Length (Metric) Exterior94.5 cm
Plug TypeEU
RefrigerantCFC-free
Shipping Weight (English)600 lb.
Shipping Weight (Metric)272 kg
TypeHigh-Performance Freezer
WarrantyWarranty varies by country, please contact us for details
Width (English) Exterior56.5 in.
Width (English) Interior52.5 in.
Width (Metric) Exterior61 cm
Width (Metric) Interior133.4 cm
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Certifications/ComplianceCE, cUL, UL
Monitoring OptionsAlarm, Recorder
No. of Doors2
Shelves8
StyleUpright
Temperature Range-30°C
Voltage230 V
Unit SizeEach