MAS™ UrichemTRAK Controls
Actual product may vary
Thermo Scientific™
MAS™ UrichemTRAK Controls
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient vial format.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionLevel
UR-MPUrichemTRAK Controls, Assayed, Bi-levelBi-level
UR22002UrichemTRAK Controls, Assayed2
UR11001UrichemTRAK Controls, Assayed1
10014127UrichemTRAK Control, Vista Vials, Assayed, Bi-levelBi-level
Catalog number UR-MP
Price (USD)
130.00
Each
Description:
UrichemTRAK Controls, Assayed, Bi-level
Level:
Bi-level
Price (USD)
130.00
Each
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient Vista™ vial format. Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.
  • Liquid, human urine matrix
  • Open and use; no equilibration required
  • Two-year open-vial stability
  • Individual levels or multi-pack configurations
  • Stable for 24 months at 2° to 8°C
  • Assayed values available for most major analyzers
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionUrichemTRAK Controls, Assayed, Bi-level
LevelBi-level
Packaging6 pack - 3 bottles of each level
Quantity15 mL
Shelf Life24 months
TypeControl
Detectable AnalytesAmylase, Calcium, Chloride, Cortisol, Creatinine, Glucose, hCG, Magnesium, Microalbumin, Osmolality, pH, Phosphorus, Potassium, Sodium, Specific Gravity, Total Protein, Urea Nitrogen, Uric Acid
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C