Search
Search
| Catalog Number | Type |
|---|---|
| UTP-TEF0.75 also known as UTP-TEF 0.75 | Tubing Pack |
| SCT120 | Chemical Trap |
| DTK120R | Chemical Trap Kit |
| DC120 | Disposable Cartridge |
| DC120A | Acid Neutralization Cartridge |
| ANC100 | Acid Neutralizing Compound |
| ANS121 | Ammonia Neutralizing Solution |
| ANT100 | Post Trap Assembly |
| MF190 | Mist Filter Replacement Cartridge |
| GCF400 | Condensation Flask |
| CC120DX also known as CC120/DX | Cart |
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.