Revco™ UxF -86°C Upright Ultra-Low Temperature Freezer, 19.4 cu. ft. (547.9L) capacity, holds 400 boxes, 115V/60Hz - FAQs
Revco™ UxF -86°C Upright Ultra-Low Temperature Freezer, 19.4 cu. ft. (547.9L) capacity, holds 400 boxes, 115V/60Hz - FAQs
View additional product information for Revco™ UxF -86°C Upright Ultra-Low Temperature Freezers - FAQs (UXF40086A)