Revco™ UxF -86°C Upright Ultra-Low Temperature Freezer, 28.8 cu. ft. (815.1L) capacity, holds 600 boxes, 208-230V/60Hz - FAQs
Revco™ UxF -86°C Upright Ultra-Low Temperature Freezer, 28.8 cu. ft. (815.1L) capacity, holds 600 boxes, 208-230V/60Hz - FAQs
View additional product information for Revco™ UxF -86°C Upright Ultra-Low Temperature Freezers - FAQs (UXF60086D)